Epic™ Basic Protection Gowns
Product Details & Documents
Latex-free, low-lint gown for use in some critical environments, and many general application areas. Breathable cloth-like fabric is comfortable and cool. Features include elastic wrists, wrap-around neck ties, and extra-long wrap-around waist ties that can be tied in front if desired.
Made from lightweight spun-bond polypropylene (SBPP) for breathability and comfort
- Non-linting design suitable for general contamination control
- Breathable material reduces heat buildup during extended wear
- Full coverage gown with long sleeves for basic protection
- Elastic cuffs for secure fit and reduced sleeve slippage
- Waist and neck ties for adjustable closure
- Meets basic hygiene and contamination control standards for non-critical environments
- Ideal for ISO Class 7 - 8 cleanrooms or general-purpose areas
- Single-use design eliminates laundering and reduces cross-contamination risk
- Economical option for visitor gowns or low-risk tasks
- Suitable for pharmaceutical, food processing, electronics assembly, and general lab use
The Epic™ Basic protection gown is designed for environments where lightweight coverage, comfort, and cost efficiency are essential. This gown provides a practical solution for general contamination control in cleanrooms, laboratories, and controlled workspaces that do not involve hazardous chemicals or high-risk exposure.
This garment is constructed from a breathable non-woven polypropylene fabric. Individually inspected for sizing, stitching, and workmanship.
Packaging: Neatly folded and cleanroom packed; 10 pieces per sealed inner polybag, 5 polybags per sealed master bag, 50 pieces per case.
Spécifications
- Taille de confection:Universal
- Color:White
- Longueur:45"
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Back fastening
- Water resistance:Very light splash
- Tensile strength:16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Weight:30 g/m²
- Tearing strength:3.4 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Cat. No.:78150-474
- Cleanroom class:10000/ISO 7
- Garment application:Cleanroom
- Pockets:Without pockets
- Fabric:Spunbound polpropylene
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:6856 g/m²/24 h
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



