Epic™ Basic Protection Gowns
Latex-free, low-lint gown for use in some critical environments, and many general application areas. Breathable cloth-like fabric is comfortable and cool. Features include elastic wrists, wrap-around neck ties, and extra-long wrap-around waist ties that can be tied in front if desired.
Made from lightweight spun-bond polypropylene (SBPP) for breathability and comfort
- Non-linting design suitable for general contamination control
- Breathable material reduces heat buildup during extended wear
- Full coverage gown with long sleeves for basic protection
- Elastic cuffs for secure fit and reduced sleeve slippage
- Waist and neck ties for adjustable closure
- Meets basic hygiene and contamination control standards for non-critical environments
- Ideal for ISO Class 7 - 8 cleanrooms or general-purpose areas
- Single-use design eliminates laundering and reduces cross-contamination risk
- Economical option for visitor gowns or low-risk tasks
- Suitable for pharmaceutical, food processing, electronics assembly, and general lab use
The Epic™ Basic protection gown is designed for environments where lightweight coverage, comfort, and cost efficiency are essential. This gown provides a practical solution for general contamination control in cleanrooms, laboratories, and controlled workspaces that do not involve hazardous chemicals or high-risk exposure.
This garment is constructed from a breathable non-woven polypropylene fabric. Individually inspected for sizing, stitching, and workmanship.
Packaging: Neatly folded and cleanroom packed; 10 pieces per sealed inner polybag, 5 polybags per sealed master bag, 50 pieces per case.
- Taille de confection:Universal
- Color:Yellow
- Longueur:45"
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Back fastening
- Water resistance:Very light splash
- Tensile strength:16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Weight:30 g/m²
- Tearing strength:3.4 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Cat. No.:78150-530
- Cleanroom class:10000/ISO 7
- Garment application:Cleanroom
- Pockets:Without pockets
- Fabric:Spunbound polpropylene
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:6856 g/m²/24 h
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



