Epic™ Basic Protection Lab Coats
Product Details & Documents
Latex-free, low-lint lab coat for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Lab coats feature plastic snaps, elastic wrists, folded collar, and three pockets.
Provides essential coverage for non-hazardous environments such as laboratories, clean areas, and general manufacturing
- Made from lightweight, breathable polypropylene non-woven fabric, ensuring comfort while maintaining contamination control
- Snap-front design for easy donning and doffing without compromising coverage
- Options for elastic wrists or open cuffs to suit different applications and glove integration
- Latex-free and lint-free design reduces risk of allergic reactions and minimizes particle generation in controlled environments
- Individually bagged and cleanroom-packed to maintain garment integrity
- Disposable design eliminates laundering costs and simplifies inventory management
Epic™ Basic protection lab coats are designed to deliver essential coverage and contamination control for non‑hazardous tasks in laboratories, controlled environments, and light manufacturing. Built from lightweight, breathable non-woven materials, these disposable coats strike a practical balance between comfort, cleanliness, and cost, making them a dependable choice for routine work, visitors, and ancillary cleanroom activities.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Certifications: Meets basic cleanroom garment guidelines for controlled environments.
Packaging: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
Specifications
- Taille de confection:3XL
- Color:White
- Seam type:Bound
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Press studs / snaps
- Tensile strength:3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Weight:30 g/m²
- Tearing strength:16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Cat. No.:78150-642
- Cleanroom class:10000/ISO 7
- Garment application:Cleanroom
- Pockets:With pockets
- Fabric:SPP
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:6856 g/m²/24 hrs.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



