Epic™ Basic Protection Lab Coats
Product Details & Documents
Latex-free, low-lint lab coat for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Lab coats feature plastic snaps, elastic wrists, folded collar, and three pockets.
Provides essential coverage for non-hazardous environments such as laboratories, clean areas, and general manufacturing
- Made from lightweight, breathable polypropylene non-woven fabric, ensuring comfort while maintaining contamination control
- Snap-front design for easy donning and doffing without compromising coverage
- Options for elastic wrists or open cuffs to suit different applications and glove integration
- Latex-free and lint-free design reduces risk of allergic reactions and minimizes particle generation in controlled environments
- Individually bagged and cleanroom-packed to maintain garment integrity
- Disposable design eliminates laundering costs and simplifies inventory management
Epic™ Basic protection lab coats are designed to deliver essential coverage and contamination control for non‑hazardous tasks in laboratories, controlled environments, and light manufacturing. Built from lightweight, breathable non-woven materials, these disposable coats strike a practical balance between comfort, cleanliness, and cost, making them a dependable choice for routine work, visitors, and ancillary cleanroom activities.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Certifications: Meets basic cleanroom garment guidelines for controlled environments.
Packaging: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
Specifications
- Taille de confection:XL
- Color:Blue
- Seam type:Bound
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Press studs / snaps
- Tensile strength:3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Weight:30 g/m²
- Tearing strength:16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Cat. No.:78150-626
- Cleanroom class:10000/ISO 7
- Garment application:Cleanroom
- Pockets:With pockets
- Fabric:SPP
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:6856 g/m²/24 hrs.



