Epic Navy Basic Protection Coveralls
Product Details & Documents
Latex free/low lint coverall for use in some critical/clean environments, and many industrial applications. Nonwoven fabric is cloth-like and cool to wear. Features include a zipper front entry and elastic wrists.
Cleanroom-compatible design: Engineered for controlled environments to minimize particle shedding
- Basic protection level: Provides essential barrier against non-hazardous particulates and light liquid splashes
- Durable, low-linting fabric: Helps maintain cleanroom integrity and reduces contamination risk
- Full-body coverage: Includes long sleeves, full-length legs, and secure front closure
- Elastic cuffs: Ensures a snug fit and reduces particle escape
- Secure closure: Snap or zipper front for quick donning and doffing
- Available sizes: Multiple size options for proper fit across personnel
Full‑body, low‑lint protection for non‑critical cleanroom tasks.
Garment is constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
The Tians Epic Basic Protection Cleanroom Coverall delivers reliable particle barrier and comfort for routine operations in controlled environments. Made from durable, low‑linting fabric, it helps maintain cleanroom integrity while supporting operator mobility and all‑day wear. Featuring secure front closure, elasticized cuffs and ankles, and optional hood/boot integration (model‑dependent), this coverall is a cost‑effective choice for pharmaceutical, electronics, and medical device applications that require basic contamination control.
Benefits:
Contamination control: Maintains cleanliness standards in ISO Class cleanrooms and controlled environments.
Operator comfort: Lightweight, breathable material supports extended wear without discomfort.
Cost efficiency: Ideal for non-critical processes, reducing unnecessary expense on higher-level garments.
Compliance support: Meets cleanroom apparel requirements for GMP and ISO standards.
Versatile use: Suitable for pharmaceutical, electronics, and medical device manufacturing.
Ball burst: 82.58 N
Flammability: Class I
Packaging: Neatly folded and packed 25 pieces per case in a sealed inner poly-bag.
Spécifications
- Taille de confection:XL
- Color:Blue
- Packaging:25/case
- Type:Wrist cuff: Blue
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Zip closure
- Water resistance:No resistance
- Tensile strength:16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
- Weight:30 g/m²
- Tearing strength:3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
- Cat. No.:78150-662
- Cleanroom class:10000/ISO 7
- Garment application:Cleanroom
- Pockets:Without pockets
- Fabric:Spunbound polypropylene
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:6856 g/m²/24 h



