VWR® Screw Cap Microtubes with Socket Caps
Product Details & Documents
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards
- High-clarity design: Crystal-clear walls for easy sample visibility
- Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
- Designed with an insertable ridge for convenient sample coding using colorful insert chips
- Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
- Sterile and non sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
- Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
: Spare caps and color insert chips also available.
Certificates
Recherche par numéro de lotSpécifications
- Cap Type:Tethered, socket
- Format:Conical base
- Sterility:Non sterile
- Volume:1.5 ml
- Color:Clear
- Packaging:Combo pack
- Maximum RCF:20000×g
- Cap Color:Clear
- Graduation:Yes



