VWR® Screw Cap Microtubes with Socket Caps
Product Details & Documents
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards
- High-clarity design: Crystal-clear walls for easy sample visibility
- Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
- Designed with an insertable ridge for convenient sample coding using colorful insert chips
- Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
- Sterile and non sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
- Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
Ordering information: Spare caps and color insert chips also available.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Cap Type:Tethered, socket
- Format:Freestanding
- Sterility:Non sterile
- Volume:0.5 ml
- Color:Clear
- Packaging:Combo pack
- Maximum RCF:20000×g
- Cap Color:Clear
- Graduation:No
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



