Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Screw Cap Microtubes with Socket Caps

VWR® Screw Cap Microtubes with Socket Caps

Clear, 2.0 ml, Clear, Freestanding, Tethered, socket, Yes, 20000×g, Combo pack, Non sterile
Catalog # 78115-716
New Product
Supplier:   VWR International
VWR® Screw Cap Microtubes with Socket Caps
Catalog # 78115-716
Supplier:  VWR International
New Product
VWR® Screw Cap Microtubes with Socket Caps
VWR® Screw Cap Microtubes with Socket Caps

Clear, 2.0 ml, Clear, Freestanding, Tethered, socket, Yes, 20000×g, Combo pack, Non sterile

Price
$112.99
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 500 Case of 5000
Product Details & Documents

VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.

  • Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards

  • High-clarity design: Crystal-clear walls for easy sample visibility
  • Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
  • Designed with an insertable ridge for convenient sample coding using colorful insert chips
  • Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
  • Sterile and non sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
  • Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications

VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.

Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.

Ordering information: Spare caps and color insert chips also available.

Documents

Spécifications
  • Cap Type:
    Tethered, socket
  • Format:
    Freestanding
  • Sterility:
    Non sterile
  • Volume:
    2.0 ml
  • Color:
    Clear
  • Packaging:
    Combo pack
  • Maximum RCF:
    20000×g
  • Cap Color:
    Clear
  • Graduation:
    Yes