VWR® Bin Liners | Avantor
VWR® Bin Liners
BioHazard bin liner, flat bottom, irradiated, Red, LDPE, 38×48", 3 mil, Individually bagged and tagged in a double bagged pack
Catalog # 78107-338
New Product
Supplier:   VWR International
VWR® Bin Liners
Catalog # 78107-338
Supplier:  VWR International
New Product
VWR® Bin Liners
VWR® Bin Liners
BioHazard bin liner, flat bottom, irradiated, Red, LDPE, 38×48", 3 mil, Individually bagged and tagged in a double bagged pack
Price
$646.58
Log in pour la disponibilité
Case of 50
Product Details & Documents
LDPE liners for use in clean and controlled aseptic environments.
  • Available non sterile or gamma irradiated
Uses include transport of product and waste disposal.
Spécifications
  • Color:
    Red
  • Désignation:
    BioHazard bin liner, flat bottom, irradiated
  • Dimensions:
    38×48"
  • Matière:
    LDPE
  • Épaisseur:
    3 mil
  • Packaging:
    Individually bagged and tagged in a double bagged pack