VWR® Bin Liners
BioHazard bin liner, flat bottom, irradiated, Red, LDPE, 38×48", 3 mil, Individually bagged and tagged in a double bagged pack
Catalog # 78107-338
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
LDPE liners for use in clean and controlled aseptic environments.
- Available non sterile or gamma irradiated
Uses include transport of product and waste disposal.
Documents
Spécifications
- Color:Red
- Désignation:BioHazard bin liner, flat bottom, irradiated
- Dimensions:38×48"
- Matière:LDPE
- Épaisseur:3 mil
- Packaging:Individually bagged and tagged in a double bagged pack