VWR® Bin Liners
BioHazard bin liner, flat bottom, Red, LDPE, 38×48", 3 mil, Double-bagged in groups of 5
Catalog # 78107-336
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
LDPE liners for use in clean and controlled aseptic environments.
- Available non sterile or gamma irradiated
Uses include transport of product and waste disposal.
Documents
Specifications
- Color:Red
- Désignation:BioHazard bin liner, flat bottom
- Dimensions:38×48"
- Matière:LDPE
- Épaisseur:3 mil
- Packaging:Double-bagged in groups of 5