Polyethylene Dropping Bottles
29 ml (1 oz.), LDPE
Catalog # 470356-535
New Product
Supplier: Dynalon
Fournisseur #: 605065-0010
Jump to:
Product Details & Documents
Economy LDPE dropper bottles.
- Translucent LDPE dropper bottles
- Sturdy white dropping cap
- Flip up spout
Dropper bottles with hinged dispenser caps for dispensing indicator solutions, dyes, and more.
Spécifications
- Capacité:29 ml (1 oz.)
- Matière:LDPE