TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
TSK-Gel® Super-Octyl, 11 nm, C8 (Octyl), 110 Å, Acier inoxydable, 2 µm, 2.00 mm, 50 mm, 0.15 - 0.20 ml/min, Slumns, Tosohoel® ReverroKyl, -
Catalog # 78049-252
New Product
Supplier: Tosoh Bioscience
Fournisseur #: 20013
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
Catalog # 78049-252
Supplier: Tosoh Bioscience
New Product
Jump to:
Product Details & Documents
Recommended for small molecular weight compounds (<10000 Da) such as peptides, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food and beverage samples.
- Provide superior resolution and speed
- Improved sensitivity
- Pressure drop is not excessive due to the monodisperse particle size distribution
TSKgel® Super-ODS, Super-Octyl and Super-Phenyl packings are bonded with, respectively, C18, C8 and phenyl functional groups. The bonded phases have a polymeric structure. An exhaustive endcapping reaction minimizes the presence of residual silanol groups.
Spécifications
- Débit:0.15 - 0.20 ml/min
- Ø int.:2.00 mm
- Granulométrie:2 µm
- Taille de pore:110 Å
- Désignation:TSK-Gel® Super-Octyl, 11 nm
- Longueur:50 mm
- Column Hardware:Acier inoxydable
- Packing Material:Slumns, Tosohoel® ReverroKyl, -
- Phase:C8 (Octyl)
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



