TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
TSK-Gel® Super-Phenyl, 11 nm, Phenyl, 110 Å, Acier inoxydable, 2 µm, 4.60 mm, 100 mm, 1 - 2.5 ml/min, Mm, e
Catalog # 78049-004
New Product
Supplier: Tosoh Bioscience
Fournisseur #: 18278
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
Catalog # 78049-004
Supplier: Tosoh Bioscience
New Product
Jump to:
Product Details & Documents
Recommended for small molecular weight compounds (<10000 Da) such as peptides, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food and beverage samples.
- Provide superior resolution and speed
- Improved sensitivity
- Pressure drop is not excessive due to the monodisperse particle size distribution
TSKgel® Super-ODS, Super-Octyl and Super-Phenyl packings are bonded with, respectively, C18, C8 and phenyl functional groups. The bonded phases have a polymeric structure. An exhaustive endcapping reaction minimizes the presence of residual silanol groups.
Spécifications
- Débit:1 - 2.5 ml/min
- Ø int.:4.60 mm
- Granulométrie:2 µm
- Taille de pore:110 Å
- Désignation:TSK-Gel® Super-Phenyl, 11 nm
- Longueur:100 mm
- Column Hardware:Acier inoxydable
- Packing Material:Mm, e
- Phase:Phenyl



