TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
TSK-Gel® Super-Phenyl, 11 nm, Phenyl, 110 Å, Acier inoxydable, 2 µm, 4.60 mm, 50 mm, 1 - 2.5 ml/min, Mm, e
Catalog # 78049-002
New Product
Supplier: Tosoh Bioscience
Fournisseur #: 18277
TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
Catalog # 78049-002
Supplier: Tosoh Bioscience
New Product
Jump to:
Product Details & Documents
Recommended for small molecular weight compounds (<10000 Da) such as peptides, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food and beverage samples.
- Provide superior resolution and speed
- Improved sensitivity
- Pressure drop is not excessive due to the monodisperse particle size distribution
TSKgel® Super-ODS, Super-Octyl and Super-Phenyl packings are bonded with, respectively, C18, C8 and phenyl functional groups. The bonded phases have a polymeric structure. An exhaustive endcapping reaction minimizes the presence of residual silanol groups.
Spécifications
- Débit:1 - 2.5 ml/min
- Ø int.:4.60 mm
- Granulométrie:2 µm
- Taille de pore:110 Å
- Désignation:TSK-Gel® Super-Phenyl, 11 nm
- Longueur:50 mm
- Column Hardware:Acier inoxydable
- Packing Material:Mm, e
- Phase:Phenyl
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



