Digital Heating/Cooling Dry Baths
À propos de cet article
Advanced dry temperature control minimizes contamination risk protecting your samples and reduces experiment time.
Advanced Peltier technology delivers rapid heat-up and cool down performance
- Precise, reliable temperature control for continuous operation
- Wide variety of interchangeable sample tube and plate blocks available
- Programmable time, temperature segments
- Integrated safety features for added protection
Significantly reduce experiment time with rapid, precise and accurate temperature management, without the need for a refrigeration system. Dry temperature control helps maintain a clean environment while the vibration free design is ideal for incubations where stable conditions are critical.
Delivery information: Includes instruction manual, power cord and two-year replacement warranty.
2 Options Available Below
Product Details & Documents
Advanced dry temperature control minimizes contamination risk protecting your samples and reduces experiment time.
Advanced Peltier technology delivers rapid heat-up and cool down performance
- Precise, reliable temperature control for continuous operation
- Wide variety of interchangeable sample tube and plate blocks available
- Programmable time, temperature segments
- Integrated safety features for added protection
Significantly reduce experiment time with rapid, precise and accurate temperature management, without the need for a refrigeration system. Dry temperature control helps maintain a clean environment while the vibration free design is ideal for incubations where stable conditions are critical.
Delivery information: Includes instruction manual, power cord and two-year replacement warranty.
Specifications for Product Family
Spécifications
- Catalog No:
- 78739-098
- 78739-100
- Model:
- DBC-05A
- DBC-06A
- Temperature setting:
- –10...+100 °C
- –10...+100 °C
- Heating rate, ambient to 100 °C:
- ≤15 min (20...100 °C)
- ≤15 min (20...100 °C)
- Cooling rate:
- ≤30 min 100...25 °C
≤30 min RT to RT–30 °C - ≤30 min 100...25 °C
≤30 min RT to RT–20 °C
- Temperature control range:
- RT–30...+100 °C
- RT −25...+100 °C
Spécifications
Product Family Options
Informations sur le produit
- DésignationCapacitéAvailabilityPrice
- Digital heating cooling dry bath0.2 ml×96/0.5 ml×54/
1.5 ml/2.0 ml×35/5 ml×24/10 ml×12/
15 ml×12/50 ml×6/2.0 ml vials×24
1 MicroplateLog in pour la disponibilitéEach$2,380.00Supplier Number: DBC-05ASpécifications:New ProductMore
Specifications
Cat. No.78739-098ModelDBC-05ATemperature setting–10...+100 °CHeating rate, ambient to 100 °C≤15 min (20...100 °C)Cooling rate≤30 min 100...25 °C
≤30 min RT to RT–30 °CTemperature control rangeRT–30...+100 °CNew Product - Digital heating cooling mini dry bath0.2 ml×32/0.5 ml×24/
1.5 ml/2.0 ml×15/5 ml×12
15 ml×6/50 ml×2/Cuvette×8Log in pour la disponibilitéEach$1,150.80Supplier Number: DBC-06ASpécifications:New ProductMore
Specifications
Cat. No.78739-100ModelDBC-06ATemperature setting–10...+100 °CHeating rate, ambient to 100 °C≤15 min (20...100 °C)Cooling rate≤30 min 100...25 °C
≤30 min RT to RT–20 °CTemperature control rangeRT −25...+100 °CNew Product
Accessories
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 12×10 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 35×1.5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 12×15 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 6×50 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 32×0.2 ml+ 10×0.5 ml+15×2.0 ml tubesBlock 32×0.2 ml+ 10×0.5 ml+15×2.0 ml tubes78739-118DBC-05ALog in pour la disponibilitéEach - $350.00
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 32×0.2 ml+10×0.5 ml+15×1.5 ml tubesBlock 32×0.2 ml+10×0.5 ml+15×1.5 ml tubes78739-116DBC-05ALog in pour la disponibilitéEach - $350.00
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 24×0.5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 2×50 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 8×cuvettes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 103×67×30 mm (Open reservior)
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 6×15 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 96×0.2 ml PCR tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 54×0.5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 32×0.2 ml+25×1.5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 20×0.5 ml+15×1.5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Lid, clear for DBCA13-15
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 12×5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 24×5 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Lid, clear for DBCA11-12
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 15×2.0 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 24×2.0 ml vials
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 35×2.0 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 32×0.2 ml tubes
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Lid, clear for DBCA1-10
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 96×0.2 ml ELISA plates
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Block 15×1.5 ml tubes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



