Security Sample Bags
Security Sample Bags
New Product
Supplier:   BURKLE INC
Security Sample Bags
Supplier:  BURKLE INC
New Product
Certificates
Security Sample Bags
3 Options Available Below
Product Details & Documents
Reliable sample labelling and safe handling are essential, whether in the laboratory, in quality assurance, or in production. In laboratories, samples and retained samples are routinely sent in security bags and recorded digitally. In industry, eight-digit numbering and tamper-proof closures ensure the reliable traceability of quality samples.
  • 8 digit consecutive numbering for clear identification and complete traceability
  • Fill-in fields for clear and structured documentation
  • Security seal reliably protects against tampering
  • Barcode 128 for fast, automated data capture
  • Tear-off tab with control number for easy handling and double security
  • Bags are made from 80% recycled material
These security bags are a sophisticated solution for safely transporting, clearly identifying, and fully tracing sensitive samples. The bags and tear-off panel feature eight-digit consecutive numbering. The tamper-proof closure and practical labelling fields meet the highest standards of documentation, security, and efficiency in sample logistics and for retained samples. The bags are also impressive in terms of sustainability, as they are made from 80% recycled material.
Thanks to the fill-in fields for name, batch, customer, location, date, and signature, each sample can be assigned in a structured and traceable manner, thereby minimizing errors. The security seal with visible void marking indicates immediately whether the bag has been opened without authorisation. The integrated barcode with 128 coding enables fast digital processing and reduces sources of error. The tear-off tab with a numbered section also facilitates logging in the system or laboratory book, ensuring full control at all times.
Product Family Options
Informations sur le produit
  • Capacité
    Épaisseur
    Matière
    W×L
    Color
    Availability
    Price
  • 500 ml
    0.12 mm
    LDPE
    145×275 mm
    Transparent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $83.38
    $83.38
    Pack of 100
       $83.38
    Supplier Number: 5344-2050
    Spécifications:
    New Product
    New Product
  • 1000 ml
    0.12 mm
    LDPE
    195×300 mm
    Transparent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $102.40
    $102.40
    Pack of 100
       $102.40
    Supplier Number: 5344-2100
    Spécifications:
    New Product
    New Product
  • 2000 ml
    0.12 mm
    LDPE
    245×370 mm
    Transparent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $127.25
    $127.25
    Pack of 100
       $127.25
    Supplier Number: 5344-2200
    Spécifications:
    New Product
    New Product