Nitrate Test Kits | Avantor
Nitrate Test Kits
New Product
Supplier:   CHEMetrics
Nitrate Test Kits
Supplier:  CHEMetrics
New Product
Certificates
Nitrate Test Kits
Nitrate Test Kits
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The NED method is widely used in the quantitative analysis of nitrite and nitrate in water samples. NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, lower detection limits and measurement ranges are possible vs. chromotropic acid kits. In addition, the nitrate measurement uses only zinc which is less toxic than cadmium, making handling and disposal safer.
  • Measurement via zinc reduction only, safer handling and disposal
    Improved sensitivity and faster analysis time
    Simplified reordering, visual refills include NED, Zn, Acidifier and buffer under single catalog code
CHEMetrics Nitrate Test Kits use the N-(1-Naphthyl) ethylenediamine (NED) method to measure nitrate in potable, surface, storm, ground, sea, and wastewater. These kits offer improved sensitivity and faster analysis time (10 vs. 13 min) compared to our current chromotropic acid-based kits.
Like our legacy chromotropic acid reagent, NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, we are now able to offer lower detection limits and measurement ranges than our chromotropic acid kits provide.
Product Family Options
Comparators
Informations sur le produit
  • Désignation
    Méthode chimique
    Range
    Utilisation
    No. of Tests
    Availability
    Price
  • 0 - 0.1 ppm
    Industrial
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.80
    $33.80
    Each
       $33.80
    Supplier Number: C-6902
    Spécifications:
    New Product
    New Product
  • 0 - 210 ppm
    Industrial
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44.54
    $44.54
    Each
       $44.54
    Supplier Number: C-6908
    Spécifications:
    New Product
    New Product
Pipette tip
Informations sur le produit
  • Désignation
    Méthode chimique
    Range
    Utilisation
    No. of Tests
    Availability
    Price
  • Pipette tip, pack 30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 30
    Pack of 30 - $10.21
    $10.21
    Pack of 30
       $10.21
    Supplier Number: A-0171
    Spécifications:
    New Product
    New Product
Informations sur le produit
  • Désignation
    Méthode chimique
    Range
    Utilisation
    No. of Tests
    Availability
    Price
  • NED
    0 - 210 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 30
    Pack of 30 - $98.04
    $98.04
    Pack of 30
       $98.04
    Supplier Number: R-6901
    Spécifications:
    New Product
    New Product
  • NED
    0 - 1.4 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $164.75
    $164.75
    Each
       $164.75
    Supplier Number: K-6901
    Spécifications:
    New Product
    New Product
  • NED
    0 - 210 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $185.65
    $185.65
    Each
       $185.65
    Supplier Number: K-6901A
    Spécifications:
    New Product
    New Product
  • NED
    0 - 55 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $137.73
    $137.73
    Each
       $137.73
    Supplier Number: K-6901D
    Spécifications:
    New Product
    New Product
  • NED
    0 - 1.5 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $110.30
    $110.30
    Each
       $110.30
    Supplier Number: K-6973
    Spécifications:
    New Product
    New Product
  • NED
    0 - 15 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $110.74
    $110.74
    Each
       $110.74
    Supplier Number: K-6983
    Spécifications:
    New Product