Starlab
Starlab
New Product
Supplier:   Science First
Starlab
Supplier:  Science First
New Product
Certificates
9 Options Available Below
Product Details & Documents
Portable immersive domes with projection and NGSS-aligned curriculum for science instruction.
  • All-in-one system combining dome, projection, computer, and curriculum
  • Portable, inflatable domes enable use in classrooms, gyms, and shared spaces
  • Immersive full-dome visuals increase attention and concept comprehension
  • NGSS-aligned astronomy, geology, and meteorology curriculum available
  • Plug-and-play setup in 20 minutes minimizes technical and instructional prep time
  • Scales from small classes to large-group programs and events
  • Supports curriculum standards across K-12
The Starlab system is designed to remove the traditional barriers between students and complex scientific concepts by transforming any suitable space into an immersive learning environment. Rather than relying on fixed facilities or specialized rooms, Starlab gives educators the flexibility to deliver visually rich science instruction in an engaging, inflatable dome, supporting a wide range of instructional settings from daily classroom use to special programs and outreach events.
Each component of Starlab is engineered to work together as a cohesive whole, reducing friction for educators and ensuring a smooth experience for learners. This approach allows institutions to standardize immersive instruction across classrooms, grade levels, and locations without requiring extensive technical expertise or infrastructure changes.
Starlab also supports instructional depth by helping educators teach concepts that are difficult to convey through textbooks or flat screens alone. Spatial relationships, large-scale systems, and time-based changes become easier for students to grasp when experienced visually and contextually. This helps bridge gaps in understanding and supports a more intuitive connection to scientific ideas.
From an operational standpoint, the system is built for repeated use over time. Its design supports shared ownership across departments or schools, making it suitable for centralized resource models. Institutions can deploy Starlab as part of regular instruction, enrichment programming, or special events while maintaining consistent quality and performance.
Ultimately, the Starlab system serves as a versatile instructional platform that adapts to different educational goals and environments. It enables schools and organizations to elevate science learning experiences without adding complexity, helping educators focus on teaching while students focus on discovery.
Ordering information: Please note: The Starlab system requires at least one dome, one projector, and one curriculum package (all sold seprately) to operate and deliver a complete instructional experience. Contact your sales representative for more information.
Packaging: All packages will ship via Freight.
Product Family Options
Dome
Product Information
  • Désignation
    No. of Students
    Taille
    Diameter
    Width
    Hauteur
    Availability
    Price
  • Inflatable learning dome with blower, 4 m
    15+ (No. of elementary students; middle and high school may be less)
    19W×19D×13' H (operating clearance)
    14' (dome body)
    14' (at the door)
    11'0" clearance
    Log in for availability
    Log in for availability
    Each
    Each - $15,959.56
    $15,959.56
    Each
       $15,959.56
    Minimum order quantity is 1
    Supplier Number: 401-4944
    Specifications:
    New Product
    New Product
  • Inflatable learning dome with blower, 5 m
    25+ (No. of elementary students; middle and high school may be less)
    25W×25D×15' H (operating clearance)
    17' (dome body)
    20' (at the door)
    13'6" clearance
    Log in for availability
    Log in for availability
    Each
    Each - $15,959.56
    $15,959.56
    Each
       $15,959.56
    Minimum order quantity is 1
    Supplier Number: 401-4945
    Specifications:
    New Product
    New Product
  • Inflatable learning dome with blower, 6 m
    38+ (No. of elementary students; middle and high school may be less)
    26W×26D×16' H (operating clearance)
    19' (dome body)
    21' (at the door)
    14'6"clearance
    Log in for availability
    Log in for availability
    Each
    Each - $16,749.56
    $16,749.56
    Each
       $16,749.56
    Minimum order quantity is 1
    Supplier Number: 401-4946
    Specifications:
    New Product
    New Product
  • Inflatable learning dome with blower, 7 m
    50+ (No. of elementary students; middle and high school may be less)
    30W×30D×18' H (operating clearance)
    23' (dome body)
    24' (at the door)
    15'6" clearance
    Log in for availability
    Log in for availability
    Each
    Each - $19,909.56
    $19,909.56
    Each
       $19,909.56
    Minimum order quantity is 1
    Supplier Number: 401-4947
    Specifications:
    New Product
    New Product
Components
Product Information
  • Désignation
    No. of Students
    Taille
    Diameter
    Width
    Hauteur
    Availability
    Price
  • Mobile case and stand for Starlab 4K projector and lens
    40H×21W×16" D
    Log in for availability
    Log in for availability
    Each
    Each - $42,661.56
    $42,661.56
    Each
       $42,661.56
    Minimum order quantity is 1
    Supplier Number: 404-1004
    Specifications:
    New Product
    New Product
  • Preconfigured laptop for Starlab system operation (Lenovo IdeaPad 5 2-in-1, 14" Full HD [1920×1080] Multi-touch IPS 16:9"), 1.43 kg (3.15 lbs)
    320.4×214.6×15.68 mm (12.61×8.44×0.61")
    Log in for availability
    Log in for availability
    Each
    Each - $3,475.98
    $3,475.98
    Each
       $3,475.98
    Minimum order quantity is 1
    Supplier Number: 404-4001
    Specifications:
    New Product
    New Product
  • Astronomy software package for small dome environments
    Log in for availability
    Log in for availability
    Each
    Each - $19,437.14
    $19,437.14
    Each
       $19,437.14
    Minimum order quantity is 1
    Supplier Number: 401-7000
    Specifications:
    New Product
    New Product
  • Earth science software package for small dome instruction
    Log in for availability
    Log in for availability
    Each
    Each - $6,163.56
    $6,163.56
    Each
       $6,163.56
    Minimum order quantity is 1
    Supplier Number: 401-7100
    Specifications:
    New Product
    New Product
  • Meteorology software package for immersive dome learning
    Log in for availability
    Log in for availability
    Each
    Each - $6,163.56
    $6,163.56
    Each
       $6,163.56
    Minimum order quantity is 1
    Supplier Number: 401-7600
    Specifications:
    New Product