Immunotag™ Porcine Complement component C7 ELISA Kit
Complement component C7 ELISA kit, 96 well plate
CA78012-304
New Product
Ready-to-use enzyme-linked immunosorbent assay kits (ELISA kits).
A Sandwich ELISA kit for the detection of Pig Complement component C7.
- Désignation:Complement component C7 ELISA kit
- Taille:96 well plate
- Cat. No.:CA78012-304
- Sensitivity:23 pg/ml
- Format:Pre-coated
- Primary antibody reactivity:Pig
- Target protein:C7
- Detection range:78 - 5000 pg/ml
- Storage temperature:Refrigerate (2...8 °C)
- Regulatory status:RUO
- Assay Type:Sandwich



