Mining Sampling Bags, Clear Poly
Mining Sampling Bags, Clear Poly
New Product
Mining Sampling Bags, Clear Poly
New Product
Mining Sampling Bags, Clear Poly
Clear poly, mining sampling bags in various sizes and mil thicknesses.
  • Suitable for field and lab applications
  • Made in Canada
Side welded seal imparts improved, with greater strength, and a resulting solid tubing bottom edge.
Product Family Options
  • Désignation
    Épaisseur
    Dimensions
    Matière
    Color
  • Poly sample bag
    4 mil
    8×14"
    Plastic
    Clear
    Case of 1,000
    Case of 1,000 - $214.35
    $214.35
    Case of 1,000
       $214.35
     : 0814391000CL
    New Product
    New Product
  • Poly sample bag
    6 mil
    8×14"
    Plastic
    Clear
    Case of 500
    Case of 500 - $146.33
    $146.33
    Case of 500
       $146.33
     : 0814500500CL
    New Product
    New Product
  • Poly sample bag
    4 mil
    10×16"
    Plastic
    Clear
    Case of 500
    Case of 500 - $147.22
    $147.22
    Case of 500
       $147.22
     : 1016390500CL
    New Product
    New Product
  • Poly sample bag
    6 mil
    10×16"
    Plastic
    Clear
    Case of 500
    Case of 500 - $200.96
    $200.96
    Case of 500
       $200.96
     : 1016500500CL
    New Product
    New Product
  • Poly sample bag
    4 mil
    12×18"
    Plastic
    Clear
    Case of 500
    Case of 500 - $202.45
    $202.45
    Case of 500
       $202.45
     : 1218390500CL
    New Product
    New Product
  • Poly sample bag
    6 mil
    12×18"
    Plastic
    Clear
    Case of 500
    Case of 500 - $247.10
    $247.10
    Case of 500
       $247.10
     : 1218500500CL
    New Product
    New Product
  • Poly sample bag
    4 mil
    16×18"
    Plastic
    Clear
    Case of 500
    Case of 500 - $281.34
    $281.34
    Case of 500
       $281.34
     : 1618390500CL
    New Product
    New Product
  • Poly sample bag
    6 mil
    16×18"
    Plastic
    Clear
    Case of 250
    Case of 250 - $186.07
    $186.07
    Case of 250
       $186.07
     : 1618500250CL
    New Product
    New Product
  • Poly sample bag
    4 mil
    18×24"
    Plastic
    Clear
    Case of 250
    Case of 250 - $190.54
    $190.54
    Case of 250
       $190.54
     : 1824380250CL
    New Product
    New Product
  • Poly sample bag
    6 mil
    18×24"
    Plastic
    Clear
    Case of 250
    Case of 250 - $265.71
    $265.71
    Case of 250
       $265.71
     : 1824500250CL
    New Product
    New Product
  • Poly sample bag
    6 mil
    14×20"
    Plastic
    Clear
    Case of 250
    Case of 250 - $186.07
    $186.07
    Case of 250
       $186.07
     : 1420500250CL