Eyepieces for Microscope
New Product
Supplier: Aven Tools
Jump to:
À propos de cet article
15 Options Available Below
Product Details & Documents
Product Family Options
01_Eye guards
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 26800B-452Spécifications:New ProductNew Product
- Supplier Number: 26800B-453Spécifications:New ProductNew Product
Informations sur le produit
- DésignationAvailabilityPrice
- Wide field 10x eyepiece for DSZ-44 and NSW microscopes, Ø 29.95 mm, FN: 23Log in pour la disponibilitéEach$442.15Supplier Number: 26800B-441Spécifications:New ProductNew Product
- Wide field 15x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 15Log in pour la disponibilitéEach$478.99Supplier Number: 26800B-442Spécifications:New ProductNew Product
- Wide field 20x eyepiece for DSZ-44, DSZ-70 and NSW microscopes, Ø 29.95 mm, FN: 11.4Log in pour la disponibilitéEach$552.68Supplier Number: 26800B-443Spécifications:New ProductNew Product
- 10x eyepiece with 10:100 mm scale reticle for SPZ microscopesLog in pour la disponibilitéEach$1,105.37Supplier Number: 26800B-455Spécifications:New ProductNew Product
- 10x eyepiece with 10:100 mm scale reticle for SPZ microscopesLog in pour la disponibilitéEach$1,142.21Supplier Number: 26800B-455-10Spécifications:New ProductNew Product
- 10x eyepiece with 5:100 mm scale reticle for SPZ microscopesLog in pour la disponibilitéEach$962.88Supplier Number: 26800B-456Spécifications:New ProductNew Product
- 10x eyepiece with 20×20, 0.5 mm squares reticle for SPZ microscopesLog in pour la disponibilitéEach$962.88Supplier Number: 26800B-457Spécifications:New ProductNew Product
- Wide field 10x eyepieces for DSZ-70 microscopes, Ø 29.95 mm, FN: 20Log in pour la disponibilitéEach$302.06Supplier Number: 26800B-445Spécifications:New ProductNew Product
- Wide field 10x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 23Log in pour la disponibilitéEach$442.15Supplier Number: 26800B-448Spécifications:New ProductNew Product
- Wide field 15x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 15Log in pour la disponibilitéEach$552.68Supplier Number: 26800B-449Spécifications:New ProductNew Product
- Wide field 20x eyepieces for SPZ microscopes, Ø 29.95 mm, FN: 11.4Log in pour la disponibilitéEach$663.22Supplier Number: 26800B-450Spécifications:New ProductNew Product
- 10x Microscope eyepiece with 10×10, 1 mm squares Scale reticle, SPZ microscopesLog in pour la disponibilitéEach$962.88Supplier Number: 26800B-458Spécifications:New ProductNew Product
- 10x eyepiece with 10:100 mm cross hair reticle, SPZ microscopesLog in pour la disponibilitéEach$962.88Supplier Number: 26800B-459Spécifications:New ProductNew Product
01_Eye guards
Informations sur le produit
- Catalog #DésignationAvailabilityPrice