Computar MLH 10× Lens | Avantor
Computar MLH 10× Lens
New Product
Supplier:   Aven Tools
Computar MLH 10× Lens
Supplier:  Aven Tools
New Product
Certificates
Computar MLH 10× Lens
2 Options Available Below
Product Details & Documents
Computar manual zoom lenses provide manual adjustment of focal length, iris and focus.
  • f/5.6 to 32C manual iris
  • 21.6 to 4.5° diagonal FOV
  • Manual zoom (10×) and focus
  • Focus: 0.15 to 0.45 m
  • C-Mount connection
For use where a variable angle of view is required. Main applications for this lens include inspection, machine vision and robotics.This system requires an adapter plate.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Macro Zoom 3 - 42× lens system
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,616.62
    $1,616.62
    Each
       $1,616.62
    Supplier Number: 26700-181
    Spécifications:
    New Product
    New Product
  • Adapter plate for Macro lens
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.85
    $175.85
    Each
       $175.85
    Supplier Number: 26700-181AP
    Spécifications:
    New Product