Adjustable Wrenches or Crescent Wrenches
Adjustable Wrenches or Crescent Wrenches
New Product
 :   Aven Tools
Adjustable Wrenches or Crescent Wrenches
New Product
Adjustable Wrenches or Crescent Wrenches
Also referred to as a crescent wrench or adjustable spanner, this adjustable wrench features auto-locking jaws that help prevent loosening or slipping.
  • Handle hole for tethering, hanging or storage
    Can be autoclaved to temperatures up to 250 °F
This very popular tool has a different jaw capacity that fits many different shapes and sizes. It is made from 304 series stainless steel with a mirror polished surface that provides excellent corrosion resistance.
Product Family Options
  • Overall Length
    Opening Size
    Weight
  • 6"
    ¹⁵/₁₆"
    0.15 lb
    Each
    Each - $269.58
    $269.58
    Each
       $269.58
     : ST8115-1004
    New Product
    New Product
  • 8"
    1¹/₈"
    0.60 lb
    Each
    Each - $342.67
    $342.67
    Each
       $342.67
     : ST8115-1006
    New Product
    New Product
  • 10"
    1⁵/₁₆"
    0.75 lb
    Each
    Each - $415.95
    $415.95
    Each
       $415.95
     : ST8115-1008
    New Product
    New Product
  • 12"
    1¹/₂"
    1.40 lb
    Each
    Each - $523.71
    $523.71
    Each
       $523.71
     : ST8115-1010