Adjustable Wrenches or Crescent Wrenches
Also referred to as a crescent wrench or adjustable spanner, this adjustable wrench features auto-locking jaws that help prevent loosening or slipping.
Handle hole for tethering, hanging or storage
Can be autoclaved to temperatures up to 250 °F
This very popular tool has a different jaw capacity that fits many different shapes and sizes. It is made from 304 series stainless steel with a mirror polished surface that provides excellent corrosion resistance.
Also referred to as a crescent wrench or adjustable spanner, this adjustable wrench features auto-locking jaws that help prevent loosening or slipping.
Handle hole for tethering, hanging or storage
Can be autoclaved to temperatures up to 250 °F
This very popular tool has a different jaw capacity that fits many different shapes and sizes. It is made from 304 series stainless steel with a mirror polished surface that provides excellent corrosion resistance.
Product Family Options
- Overall LengthOpening SizeWeight
- : ST8115-1004New ProductNew Product
- : ST8115-1006New ProductNew Product
- : ST8115-1008New ProductNew Product
- : ST8115-1010New ProductNew Product