All Purpose Standard Scissors, 8"
8" all purpose standard scissors, 3³/₄", 0.25 lbs.
Catalog # 78620-980
New Product
Supplier: Aven Tools
Fournisseur #: 11023
Jump to:
Product Details & Documents
The most common scissor for a variety of applications.
- Fully polished stainless steel provides excellent corrosion resistance
- Durable stainless steel blade
Spécifications
- Désignation:8" all purpose standard scissors
- Blade Length:3³/₄"
- Weight:0.25 lbs.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



