Technik™ Cutter
New Product
Supplier:   Aven Tools
Technik™ Cutter
Supplier:  Aven Tools
New Product
Certificates
3 Options Available Below
Product Details & Documents
End cutters are used to cut close to the surface of your material, leaving a smooth surface behind.
  • Manufactured from 410 stainless steel
    Box Joint construction provides superior jaw alignment and durability
    Induction hardened jaws
    Dual leaf springs
    ESD Safe cushioned grips
Specifications for Product Family
Spécifications
  • Catalog No:
  • 78620-528
  • 78620-596
  • 78621-052
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
Spécifications
Product Family Options
Informations sur le produit
  • Matière
    Cutting Capacity
    Blade Length
    Overall Length
    Jaw Width
    Jaw Type
    Availability
    Price
  • Stainless Steel 410
    0.2 - 1.25mm soft copper wire
    17.5 mm
    114 mm
    11.85 mm
    End cutter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.31
    $39.31
    Each
       $39.31
    Supplier Number: 10327
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78620-528
    Disposable / reusable
    Reusable
    New Product
  • Stainless Steel 410
    0.2 - 1.25mm soft copper wire
    114 mm
    11.85 mm
    Oblique cutter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.31
    $39.31
    Each
       $39.31
    Supplier Number: 10328
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78620-596
    Disposable / reusable
    Reusable
    New Product
  • Stainless Steel 410
    0.1 - 1.25mm soft copper wire
    10.3 mm
    110 mm
    9.5 mm
    Tapered miniature cutter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.31
    $39.31
    Each
       $39.31
    Supplier Number: 10331
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78621-052
    Disposable / reusable
    Reusable