Professional Automatic Wire Strippers
Professional Automatic Wire Strippers
New Product
Supplier:   Aven Tools
Professional Automatic Wire Strippers
Supplier:  Aven Tools
New Product
Certificates
Professional Automatic Wire Strippers
3 Options Available Below
Product Details & Documents
Professional grade wire stripper with a spring-loaded handle and a comfort grip, designed for easy wire stripping. Includes a built-in safety shield.
  • Quick and easy wire stripping
    Professional-quality tool for fast & consistent wire stripping
    Easy to use - simply insert wire and squeeze handle
    Spring mechanism automatically holds wire, cuts & strips insulation clean leaving bare wire
    Non-slip grips, steel mechanism - great for home, auto, A/V wiring
    Built-in peg hole for storing on a pegboard, and can be used to hang from a tool belt
    Tempered high-carbon steel blades are designed for thousands of cuts
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Cutting Capacity
    Handle Type
    Weight
    Availability
    Price
  • Professional automatic wire stripper, Range: 24 - 12 AWG
    Carbon steel
    24/16/14/12 AWG wire stripping
    Spring loaded
    0.4536 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.02
    $67.02
    Each
       $67.02
    Supplier Number: 10105A
    Spécifications:
    New Product
    New Product
  • Professional automatic wire stripper, Range: 18 - 8 AWG
    Carbon steel
    18/14/10/8 AWG wire stripping
    Spring loaded
    0.4536 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.02
    $67.02
    Each
       $67.02
    Supplier Number: 10105B
    Spécifications:
    New Product
    New Product
  • Professional automatic wire stripper, Range: 16 - 8 AWG
    Carbon steel
    16/14/10/8 AWG wire stripping
    Spring loaded
    0.4536 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.02
    $67.02
    Each
       $67.02
    Supplier Number: 10105D
    Spécifications:
    New Product
    New Product