OptiXTRACT Sample Recovery Aid | Avantor
OptiXTRACT Sample Recovery Aid
Main assembly holder, OptiXTRACT 25 mm, Post-density gradient centrifugation sample recovery workflows
Catalog # CA78617-174
New Product
Supplier:   Beckman Coulter
Fournisseur #:  D17685
OptiXTRACT Sample Recovery Aid
Catalog # CA78617-174
Supplier:  Beckman Coulter
New Product
OptiXTRACT Sample Recovery Aid
OptiXTRACT Sample Recovery Aid
Main assembly holder, OptiXTRACT 25 mm, Post-density gradient centrifugation sample recovery workflows
Price
$4,732.00
Log in pour la disponibilité
Each
Product Details & Documents
The OptiXTRACT Sample Recovery Aid brings clarity and control to density gradient band extraction.
The tube mount, guided syringe rail, LED illumination and millimetre graduations help you clearly see your bands for accurate and consistent sample recovery. The result is a safe, reproducible recovery of multiple bands across the tubes with less guesswork.
Spécifications
  • Désignation:
    Main assembly holder, OptiXTRACT 25 mm
  • Compatible with:
    Post-density gradient centrifugation sample recovery workflows
  • Cat. No.:
    CA78617-174
  • Functions:
    Band extraction tool for density gradient ultracentrifugation