OptiXTRACT Adapter Kit | Avantor
OptiXTRACT Adapter Kit
Startup adapter kit OptiXTRACT, includes 50 adapter pieces, Designed specifically for proper placement of the syringe-needle assembly on the OptiXTRACT tower
Catalog # CA78617-180
New Product
Supplier:   Beckman Coulter
Fournisseur #:  D18845
OptiXTRACT Adapter Kit
Catalog # CA78617-180
Supplier:  Beckman Coulter
New Product
OptiXTRACT Adapter Kit
OptiXTRACT Adapter Kit
Startup adapter kit OptiXTRACT, includes 50 adapter pieces, Designed specifically for proper placement of the syringe-needle assembly on the OptiXTRACT tower
Price
$622.00
Log in pour la disponibilité
Each
Product Details & Documents
OptiXTRACT Adapter kit consists of 50 pieces of adapters needed to connect the syringe to the needle and placement of the assembly on the OptiXTRACT tower.
Spécifications
  • Désignation:
    Startup adapter kit OptiXTRACT, includes 50 adapter pieces
  • Compatible with:
    Designed specifically for proper placement of the syringe-needle assembly on the OptiXTRACT tower
  • Cat. No.:
    CA78617-180
  • Functions:
    Enables secure connection of the syringe to the needle for use with the OptiXTRACT system