E-Arm Focus Mount
E-Arm Focus Mount
E-arm focus mount, 3 lbs.
Catalog # 78620-960
New Product
Supplier:   Aven Tools
Supplier #:  26800B-518
E-Arm Focus Mount
Catalog # 78620-960
Supplier:  Aven Tools
New Product
E-Arm Focus Mount
E-arm focus mount, 3 lbs.
Price
$522.70
Log in for availability
Each
Product Details & Documents
Aven’s E-Arm Focus mount with 74 mm diameter ring with 15.875 mm (5/8") adapter can be mounted on any of our boom stands or articulating arm stands. Will accept SSZ, SPZ DSZ and SPZ series microscopes.  
  • Rack and Pinion mechanism with coarse focus adjustment
  • 50 mm focusing distance in vertical direction
Specifications
  • Désignation:
    E-arm focus mount, 3 lbs.