Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sharp Tweezers | Avantor

Sharp Tweezers

Sharp tweezers, Stainless steel, 80 mm, Medium sharp, serrated, 22.7 g
Catalog # 78620-878
New Product
Supplier:   Aven Tools
Fournisseur #:  18483
Sharp Tweezers
Catalog # 78620-878
Supplier:  Aven Tools
New Product
Sharp Tweezers

Sharp tweezers, Stainless steel, 80 mm, Medium sharp, serrated, 22.7 g

Price
$8.31
Log in pour la disponibilité
Each
Product Details & Documents

Made of stainless steel, these medium-sharp, serrated-tip tweezers are ideal for bending, separating and removing small items.

  • Stainless steel finish provides excellent corrosion resistance

    Ideal for bending, separating and removing small items

    Blunt sharp tip is helpful when working in confined spaces

Spécifications
  • Désignation:
    Sharp tweezers
  • Longueur:
    80 mm
  • Matière:
    Stainless steel
  • Tip Style:
    Medium sharp, serrated
  • Weight:
    22.7 g