Wright Stain Buffers
New Product
Supplier: CHAPTEC INC CA
About this item
Wright Stain Buffer is a phosphate-based solution buffered to pH 6.80. It is designed for use with the Wright Stain and Wright-Giemsa Stain hematology stains.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Wright Stain Buffer is a phosphate-based solution buffered to pH 6.80. It is designed for use with the Wright Stain and Wright-Giemsa Stain hematology stains.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA78612-136
- CA78612-250
- CA78612-226
- CA78612-144
- CA78612-146
- CAS No.:
- 7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)
- 7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)
- 7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)
- 7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)
- 7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)
Specifications
Product Family Options
Product Information
- DésignationUtilisationVolumeType de conditionnementAvailabilityPrice
- Supplier Number: TUY1010Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.CA78612-136CAS No.7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)New Product - Supplier Number: TUY1010-1Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.CA78612-250CAS No.7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)New Product - Supplier Number: TUY1010-10Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.CA78612-226CAS No.7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)New Product - Supplier Number: TUY1010-20Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.CA78612-144CAS No.7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)New Product - Supplier Number: TUY1010-500Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.CA78612-146CAS No.7778-77-0 (0.1 - 1.0%); 7558-79-4 (0.1 - 1.0%)New Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



