CE Pre-Prep Box Rack
Stainless steel pre-prep box rack
Catalog # 78496-758
New Product
Supplier: Vileda Professional - FHP
Fournisseur #: 179010
Jump to:
Product Details & Documents
Pre-Prep Box Holder is an easy addition to existing trolleys.
- Specific to cleanroom cleaning
- Fully autoclavable and chemical-resistant
- High durability and stability
The CE Pre-Prep Box Rack is designed to add compatibility of two pre-prep boxes to the CE compact or Double Bucket Trolley.
Spécifications
- Désignation:Stainless steel pre-prep box rack