Epic™ Solid Polyethylene Shoe Covers
Product Details & Documents
Lint-free disposable cleanroom shoe cover with heat sealed seams. For use in critical and low particulate clean environments, or any general industrial application. Fabric is suitable for exposure to bloodborne pathogens, some hazardous liquids, and where liquid splash is a concern.
- Made from solid polyethylene film, providing a strong, waterproof barrier
- Tear-resistant and puncture-resistant for reliable protection in demanding conditions
- Maintains integrity even in wet or chemical-prone areas
- Smooth surface prevents particle shedding, ensuring compliance with ISO cleanroom standards
- Suitable for environments requiring strict hygiene and particulate control
- Elastic opening ensures a secure fit over various shoe sizes; easy to don and remove without compromising coverage
- Zero-lint design minimizes contamination risk, maintaining controlled environment standards
- Eliminates laundering costs and simplifies inventory management
- Ideal for pharmaceutical, electronics, biotech, food processing, and other contamination-sensitive industries
- Coefficient of friction: 0.57 μs (static)
Epic™ Solid Polyethylene Covers are engineered for environments that demand maximum contamination control, liquid resistance, and operator safety. These covers are crafted from a solid polyethylene film, offering a smooth, impermeable barrier that ensures superior protection in cleanrooms, laboratories, and controlled manufacturing areas.
Fabric is constructed from a proprietary polyethylene blend, and is embossed for additional skid resistance. 5 mil material thickness, seamless bottom, and welded seams. Individually inspected for sizing, welding, and workmanship.
: Neatly cleanroom packed; 100 pieces per sealed inner polybag, 3 polybags per sealed master bag, 300 pieces per case.
Specifications
- Taille de confection:Universal
- Color:Blue
- Cat. No.:78150-810
- Cleanroom class:100/ISO 5
- Garment application:Cleanroom
- Gender:Unisex
- Fabric:Polyethylene
- Particle shedding (Helmke Drum):Level II
- Weight:42 g/m²