Epic™ Polypropylene Covers with Plain Bottom
À propos de cet article
Disposable shoe cover trimmed in latex-free elastic for an adjustable snug fit. Machine welded seams feature encapsulated elastic to reduce particulation and increase durability. Lightweight yet durable and breathable.
- 100% spunbonded polypropylene material is lightweight, breathable, and non-linting
- Breathable material minimizes heat buildup during extended use
- Smooth sole without skid coating for environments where traction enhancement is not required
- Latex-free elastic opening for secure fit around shoes or boots
- Single-use design for contamination control
- Low particulate generation; suitable for controlled environments
- Cleanroom-packed configurations for ISO Class compliance
- Disposable design eliminates laundering and reduces maintenance costs
- Ideal for cleanrooms, labs, food processing, and healthcare settings
Epic™ polypropylene shoe covers are designed for contamination control in cleanrooms, laboratories, and controlled environments where particulate management is critical. Constructed from 100% virgin spunbonded polypropylene, these covers are lightweight, breathable, and non-linting, ensuring minimal particle generation while maintaining wearer comfort during extended use. The plain bottom design offers a smooth sole without additional coatings, making it ideal for areas where enhanced traction is unnecessary or where skid-resistant materials could interfere with flooring integrity. This simplicity supports cost efficiency while maintaining essential protective performance.
100% virgin spunbonded polypropylene low-linting fabric. Machine welded seam turned inside of cover. Individually inspected for sizing, welding, and workmanship.
Packaging: Cleanroom packed 100 pieces per sealed inner polybag, 3 polybags per sealed master bag, 300 pieces per case.
4 Options Available Below
Product Details & Documents
Disposable shoe cover trimmed in latex-free elastic for an adjustable snug fit. Machine welded seams feature encapsulated elastic to reduce particulation and increase durability. Lightweight yet durable and breathable.
- 100% spunbonded polypropylene material is lightweight, breathable, and non-linting
- Breathable material minimizes heat buildup during extended use
- Smooth sole without skid coating for environments where traction enhancement is not required
- Latex-free elastic opening for secure fit around shoes or boots
- Single-use design for contamination control
- Low particulate generation; suitable for controlled environments
- Cleanroom-packed configurations for ISO Class compliance
- Disposable design eliminates laundering and reduces maintenance costs
- Ideal for cleanrooms, labs, food processing, and healthcare settings
Epic™ polypropylene shoe covers are designed for contamination control in cleanrooms, laboratories, and controlled environments where particulate management is critical. Constructed from 100% virgin spunbonded polypropylene, these covers are lightweight, breathable, and non-linting, ensuring minimal particle generation while maintaining wearer comfort during extended use. The plain bottom design offers a smooth sole without additional coatings, making it ideal for areas where enhanced traction is unnecessary or where skid-resistant materials could interfere with flooring integrity. This simplicity supports cost efficiency while maintaining essential protective performance.
100% virgin spunbonded polypropylene low-linting fabric. Machine welded seam turned inside of cover. Individually inspected for sizing, welding, and workmanship.
Packaging: Cleanroom packed 100 pieces per sealed inner polybag, 3 polybags per sealed master bag, 300 pieces per case.
Specifications for Product Family
Spécifications
- Catalog No:
- 78150-682
- 78150-734
- 78150-706
- 78150-632
- Fabric:
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
Spécifications
Product Family Options
Informations sur le produit
- ColorTaille de confectionAvailabilityPrice
- Supplier Number: 514783-2XLSpécifications:New Product
More
Specifications
Cat. No.78150-682FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroomParticle shedding (Helmke Drum)Level IINew Product - Supplier Number: 514783-LSpécifications:New Product
More
Specifications
Cat. No.78150-734FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroomParticle shedding (Helmke Drum)Level IINew Product - Supplier Number: 514783-XLSpécifications:New Product
More
Specifications
Cat. No.78150-706FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroomParticle shedding (Helmke Drum)Level IINew Product - Supplier Number: 514883-LSpécifications:New Product
More
Specifications
Cat. No.78150-632FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroomParticle shedding (Helmke Drum)Level IINew Product