Epic™ Polypropylene Covers with Plain Bottom | Avantor
Epic™ Polypropylene Covers with Plain Bottom
New Product
Epic™ Polypropylene Covers with Plain Bottom
New Product
Certificates
Epic™ Polypropylene Covers with Plain Bottom
Epic™ Polypropylene Covers with Plain Bottom
4 Options Available Below
Product Details & Documents
Disposable shoe cover trimmed in latex-free elastic for an adjustable snug fit. Machine welded seams feature encapsulated elastic to reduce particulation and increase durability. Lightweight yet durable and breathable.
  • 100% spunbonded polypropylene material is lightweight, breathable, and non-linting
  • Breathable material minimizes heat buildup during extended use
  • Smooth sole without skid coating for environments where traction enhancement is not required
  • Latex-free elastic opening for secure fit around shoes or boots
  • Single-use design for contamination control
  • Low particulate generation; suitable for controlled environments
  • Cleanroom-packed configurations for ISO Class compliance
  • Disposable design eliminates laundering and reduces maintenance costs
  • Ideal for cleanrooms, labs, food processing, and healthcare settings
Epic™ polypropylene shoe covers are designed for contamination control in cleanrooms, laboratories, and controlled environments where particulate management is critical. Constructed from 100% virgin spunbonded polypropylene, these covers are lightweight, breathable, and non-linting, ensuring minimal particle generation while maintaining wearer comfort during extended use. The plain bottom design offers a smooth sole without additional coatings, making it ideal for areas where enhanced traction is unnecessary or where skid-resistant materials could interfere with flooring integrity. This simplicity supports cost efficiency while maintaining essential protective performance.
100% virgin spunbonded polypropylene low-linting fabric. Machine welded seam turned inside of cover. Individually inspected for sizing, welding, and workmanship.
Packaging: Cleanroom packed 100 pieces per sealed inner polybag, 3 polybags per sealed master bag, 300 pieces per case.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 78150-682
  • 78150-734
  • 78150-706
  • 78150-632
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
Spécifications
Product Family Options
Informations sur le produit
  • Color
    Taille de confection
    Availability
    Price
  • Blue
    XXL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 300
    Case of 300 - $64.72
    $64.72
    Case of 300
       $64.72
    Supplier Number: 514783-2XL
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-682
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    New Product
  • Blue
    L
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 300
    Case of 300 - $62.13
    $62.13
    Case of 300
       $62.13
    Supplier Number: 514783-L
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-734
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    New Product
  • Blue
    XL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 300
    Case of 300 - $62.13
    $62.13
    Case of 300
       $62.13
    Supplier Number: 514783-XL
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-706
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    New Product
  • White
    Universal
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 300
    Case of 300 - $62.13
    $62.13
    Case of 300
       $62.13
    Supplier Number: 514883-L
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-632
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II