Epic™ Basic Protection Lab Coats
Latex-free, low-lint lab coat for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Lab coats feature plastic snaps, elastic wrists, folded collar, and three pockets.
Provides essential coverage for non-hazardous environments such as laboratories, clean areas, and general manufacturing
- Made from lightweight, breathable polypropylene non-woven fabric, ensuring comfort while maintaining contamination control
- Snap-front design for easy donning and doffing without compromising coverage
- Options for elastic wrists or open cuffs to suit different applications and glove integration
- Latex-free and lint-free design reduces risk of allergic reactions and minimizes particle generation in controlled environments
- Individually bagged and cleanroom-packed to maintain garment integrity
- Disposable design eliminates laundering costs and simplifies inventory management
Epic™ Basic protection lab coats are designed to deliver essential coverage and contamination control for non‑hazardous tasks in laboratories, controlled environments, and light manufacturing. Built from lightweight, breathable non-woven materials, these disposable coats strike a practical balance between comfort, cleanliness, and cost, making them a dependable choice for routine work, visitors, and ancillary cleanroom activities.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
: Meets basic cleanroom garment guidelines for controlled environments.
: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
Latex-free, low-lint lab coat for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Lab coats feature plastic snaps, elastic wrists, folded collar, and three pockets.
Provides essential coverage for non-hazardous environments such as laboratories, clean areas, and general manufacturing
- Made from lightweight, breathable polypropylene non-woven fabric, ensuring comfort while maintaining contamination control
- Snap-front design for easy donning and doffing without compromising coverage
- Options for elastic wrists or open cuffs to suit different applications and glove integration
- Latex-free and lint-free design reduces risk of allergic reactions and minimizes particle generation in controlled environments
- Individually bagged and cleanroom-packed to maintain garment integrity
- Disposable design eliminates laundering costs and simplifies inventory management
Epic™ Basic protection lab coats are designed to deliver essential coverage and contamination control for non‑hazardous tasks in laboratories, controlled environments, and light manufacturing. Built from lightweight, breathable non-woven materials, these disposable coats strike a practical balance between comfort, cleanliness, and cost, making them a dependable choice for routine work, visitors, and ancillary cleanroom activities.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
: Meets basic cleanroom garment guidelines for controlled environments.
: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
- Catalog No:
- 78150-604
- 78150-642
- 78150-782
- 78150-744
- 78150-740
- 78150-626
- 78150-536
- 78150-644
- 78150-784
- Fabric:
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Pockets:
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Water vapor transmission rate:
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- 6856 g/m²/24 hrs.
- Tensile strength:
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Tearing strength:
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Weight:
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
Product Family Options
Standard lab coat with elastic cuffs
- ColorTaille de confection
- : 843881-2XLNew Product
Specifications
Cat. No.78150-604FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843881-3XLNew Product
Specifications
Cat. No.78150-642FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843881-LNew Product
Specifications
Cat. No.78150-782FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843881-XLNew Product
Specifications
Cat. No.78150-744FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product
Standard lab coat with knit cuffs
- ColorTaille de confection
- : 843785-SNew Product
Specifications
Cat. No.78150-740FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843785-XLNew Product
Specifications
Cat. No.78150-626FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843785-2XLNew Product
Specifications
Cat. No.78150-536FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843785-LNew Product
Specifications
Cat. No.78150-644FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - : 843785-MNew Product
Specifications
Cat. No.78150-784FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWith pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hrs.Tensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



