Epic™ Basic Protection Cleanroom Frocks
À propos de cet article
Latex-free, low-lint frock for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Features include plastic snaps, knit wrists, and knit collar. Frocks are designed without pockets.
- Constructed from lightweight, breathable polypropylene non-woven fabric, ensuring comfort and reducing lint generation
- Snap-front design for easy gowning and removal while maintaining coverage integrity
- Elastic wrists improve glove interface
- Latex-free and lint-free design reduces allergy risk and minimizes particle shedding in cleanroom environments
- Individually bagged and cleanroom-packed to maintain garment integrity during storage and handling
- Helps prevent particulate transfer from personnel to sensitive environments, supporting GMP and ISO cleanroom protocols
- Disposable construction eliminates laundering costs and simplifies inventory management
- Ideal for ISO Class 7–8 cleanrooms, support areas, and non-critical manufacturing processes
The Epic™ Basic Protection Cleanroom Frock is designed to provide reliable coverage and contamination control for controlled environments where exposure risks are minimal but cleanliness standards remain critical. This garment offers a practical solution for cleanroom support areas, ISO Class 7–8 zones, and non-hazardous tasks, balancing comfort, compliance, and cost efficiency.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Certifications: Aligns with basic cleanroom garment guidelines for controlled environments.
Packaging: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
8 Options Available Below
Product Details & Documents
Latex-free, low-lint frock for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Features include plastic snaps, knit wrists, and knit collar. Frocks are designed without pockets.
- Constructed from lightweight, breathable polypropylene non-woven fabric, ensuring comfort and reducing lint generation
- Snap-front design for easy gowning and removal while maintaining coverage integrity
- Elastic wrists improve glove interface
- Latex-free and lint-free design reduces allergy risk and minimizes particle shedding in cleanroom environments
- Individually bagged and cleanroom-packed to maintain garment integrity during storage and handling
- Helps prevent particulate transfer from personnel to sensitive environments, supporting GMP and ISO cleanroom protocols
- Disposable construction eliminates laundering costs and simplifies inventory management
- Ideal for ISO Class 7–8 cleanrooms, support areas, and non-critical manufacturing processes
The Epic™ Basic Protection Cleanroom Frock is designed to provide reliable coverage and contamination control for controlled environments where exposure risks are minimal but cleanliness standards remain critical. This garment offers a practical solution for cleanroom support areas, ISO Class 7–8 zones, and non-hazardous tasks, balancing comfort, compliance, and cost efficiency.
These garments are constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Certifications: Aligns with basic cleanroom garment guidelines for controlled environments.
Packaging: Neatly folded and cleanroom packed; 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
Specifications for Product Family
Spécifications
- Catalog No:
- 78150-702
- 78150-566
- 78150-704
- 78150-630
- 78150-776
- 78150-494
- 78150-614
- 78150-524
- Fabric:
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- SPP
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Water vapor transmission rate:
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- Tensile strength:
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- 3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)
- Tearing strength:
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- 16 lbs./in. (machine direction), 11 lbs./in. (cross direction)
- Weight:
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
Spécifications
Product Family Options
Informations sur le produit
- ColorTaille de confectionAvailabilityPrice
- Supplier Number: 843885NP-LSpécifications:New Product
More
Specifications
Cat. No.78150-702FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-2XLSpécifications:New Product
More
Specifications
Cat. No.78150-566FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-3XLSpécifications:New Product
More
Specifications
Cat. No.78150-704FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-4XLSpécifications:New Product
More
Specifications
Cat. No.78150-630FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-5XLSpécifications:New Product
More
Specifications
Cat. No.78150-776FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-MSpécifications:New Product
More
Specifications
Cat. No.78150-494FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-SSpécifications:New Product
More
Specifications
Cat. No.78150-614FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product - Supplier Number: 843885NP-XLSpécifications:New Product
More
Specifications
Cat. No.78150-524FabricSPPClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongSeam typeBoundGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater vapor transmission rate6856 g/m²/24 hTensile strength3.5 lbs./in. (machine direction), 7.1 lbs./in. (cross direction)Tearing strength16 lbs./in. (machine direction), 11 lbs./in. (cross direction)Weight30 g/m²New Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



