Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Epic Navy Basic Protection Coveralls

New Product
Epic Navy Basic Protection Coveralls
New Product
Certificates
Epic Navy Basic Protection Coveralls

4 Options Available Below

Product Details & Documents

Latex free/low lint coverall for use in some critical/clean environments, and many industrial applications. Nonwoven fabric is cloth-like and cool to wear. Features include a zipper front entry and elastic wrists.

  • Cleanroom-compatible design: Engineered for controlled environments to minimize particle shedding

  • Basic protection level: Provides essential barrier against non-hazardous particulates and light liquid splashes
  • Durable, low-linting fabric: Helps maintain cleanroom integrity and reduces contamination risk
  • Full-body coverage: Includes long sleeves, full-length legs, and secure front closure
  • Elastic cuffs: Ensures a snug fit and reduces particle escape
  • Secure closure: Snap or zipper front for quick donning and doffing
  • Available sizes: Multiple size options for proper fit across personnel

Full‑body, low‑lint protection for non‑critical cleanroom tasks.

Garment is constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.

The Tians Epic Basic Protection Cleanroom Coverall delivers reliable particle barrier and comfort for routine operations in controlled environments. Made from durable, low‑linting fabric, it helps maintain cleanroom integrity while supporting operator mobility and all‑day wear. Featuring secure front closure, elasticized cuffs and ankles, and optional hood/boot integration (model‑dependent), this coverall is a cost‑effective choice for pharmaceutical, electronics, and medical device applications that require basic contamination control.

Benefits:
Contamination control: Maintains cleanliness standards in ISO Class cleanrooms and controlled environments.
Operator comfort: Lightweight, breathable material supports extended wear without discomfort.
Cost efficiency: Ideal for non-critical processes, reducing unnecessary expense on higher-level garments.
Compliance support: Meets cleanroom apparel requirements for GMP and ISO standards.
Versatile use: Suitable for pharmaceutical, electronics, and medical device manufacturing.

Ball burst: 82.58 N

Flammability: Class I

 : Neatly folded and packed 25 pieces per case in a sealed inner poly-bag.

Specifications for Product Family

Spécifications

  • Catalog No:
  • 78150-592
  • 78150-674
  • 78150-574
  • 78150-662
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • No resistance
  • No resistance
  • No resistance
  • No resistance
  • Water vapor transmission rate:
  • 6856 g/m²/24 h
  • 6856 g/m²/24 h
  • 6856 g/m²/24 h
  • 6856 g/m²/24 h
  • Tensile strength:
  • 16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
  • 16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
  • 16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
  • 16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
  • Tearing strength:
  • 3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
  • 3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
  • 3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
  • 3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
  • Weight:
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²

Spécifications

Product Family Options

Informations sur le produit

  • Type
    Color
    Taille de confection
    Packaging
    Availability
    Price
  • Wrist cuff: Elastic
    Blue
    2XL
    25/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - 88,01 $
    88,01 $
    Case of 25
       88,01 $

    Supplier Number: 210681-2XL
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-592
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    No resistance
    Water vapor transmission rate
    6856 g/m²/24 h
    Tensile strength
    16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
    Tearing strength
    3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
    Weight
    30 g/m²
    New Product
  • Wrist cuff: Elastic
    Blue
    3XL
    25/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - 88,01 $
    88,01 $
    Case of 25
       88,01 $

    Supplier Number: 210681-3XL
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-674
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    No resistance
    Water vapor transmission rate
    6856 g/m²/24 h
    Tensile strength
    16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
    Tearing strength
    3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
    Weight
    30 g/m²
    New Product
  • Wrist cuff: Elastic
    Blue
    L
    25/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - 88,01 $
    88,01 $
    Case of 25
       88,01 $

    Supplier Number: 210681-L
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-574
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    No resistance
    Water vapor transmission rate
    6856 g/m²/24 h
    Tensile strength
    16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
    Tearing strength
    3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
    Weight
    30 g/m²
    New Product
  • Wrist cuff: Blue
    Blue
    XL
    25/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - 88,01 $
    88,01 $
    Case of 25
       88,01 $

    Supplier Number: 210681-XL
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78150-662
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    No resistance
    Water vapor transmission rate
    6856 g/m²/24 h
    Tensile strength
    16 lbs./in (Machine direction); 11 lbs./in (Cross direction)"
    Tearing strength
    3.5 lbs./in (Machine direction); 7.1 lbs./in (Cross direction)
    Weight
    30 g/m²
    New Product