Monarch® Buffer BZ | Avantor
Monarch® Buffer BZ
Monarch® Buffer BZ, 168 ml
Catalog # CA77938-754
New Product
Fournisseur #:  T1114L
Monarch® Buffer BZ
Catalog # CA77938-754
New Product
CAS Number:  
Monarch® Buffer BZ
Monarch® Buffer BZ
Monarch® Buffer BZ, 168 ml
CAS Number:  
Price
$241.00
Log in pour la disponibilité
Each(1/Itm/Bx)
La quantité minimale de commande est de 1
Product Details & Documents
Monarch Buffer BZ is included in Monarch nucleic acid purification kits, and also offered as a standalone buffer for convenience.
  • A component of the Monarch DNA and RNA purification kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
  • Supplied as a concentrate, add appropriate alcohol before use
This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. In plasmid miniprep applications, it is used as the first wash buffer to remove contaminants such as cellular components. For DNA cleanup applications, it is utilized to dilute DNA samples and facilitate binding to the Monarch spin columns.
Spécifications
Spécifications
  • Désignation:
    Monarch® Buffer BZ
  • Taille:
    168 ml