Monarch® Buffer WZ | Avantor
Monarch® Buffer WZ
Monarch® Buffer WZ, 26 ml
Catalog # CA77938-756
New Product
Fournisseur #:  T1115L
Monarch® Buffer WZ
Catalog # CA77938-756
New Product
CAS Number:  
Monarch® Buffer WZ
Monarch® Buffer WZ
Monarch® Buffer WZ, 26 ml
CAS Number:  
Price
$53.00
Log in pour la disponibilité
Each(1/Itm/Bx)
La quantité minimale de commande est de 1
Product Details & Documents
Monarch Buffer WZ is included in Monarch nucleic acid purification kits, and also offered as a standalone buffer for convenience.
  • A component of the Monarch DNA and RNA purification kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
  • Supplied as a concentrate, add appropriate alcohol before use
This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. This is designed as a wash buffer to remove any trace buffer components and salts.
Spécifications
Spécifications
  • Désignation:
    Monarch® Buffer WZ
  • Taille:
    26 ml
Spécifications