Monarch® Buffer BY | Avantor
Monarch® Buffer BY
Monarch® Buffer BY, 230 ml
Catalog # CA77938-758
New Product
Fournisseur #:  T1121L
Monarch® Buffer BY
Catalog # CA77938-758
New Product
CAS Number:  
Monarch® Buffer BY
Monarch® Buffer BY
Monarch® Buffer BY, 230 ml
CAS Number:  
Price
$210.00
Log in pour la disponibilité
Each(1/Itm/Bx)
La quantité minimale de commande est de 1
Product Details & Documents
Monarch Buffer BY is included in Monarch nucleic acid purification kits, and also offered as a standalone buffer for convenience.
  • A component of the Monarch DNA and RNA purification kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. This buffer is used to dissolve the agarose containing the target DNA and create an optimal condition for DNA to effectively bind to the spin columns.
Spécifications
Spécifications
  • Désignation:
    Monarch® Buffer BY
  • Taille:
    230 ml
Spécifications