Monarch® Buffer B3
Monarch® Buffer B3, 110 ml
CA77938-752
New Product
: T1113L
Monarch Buffer B3 is included in Monarch nucleic acid purification kits, and also offered as a standalone buffer for convenience.
- A component of the Monarch DNA and RNA purification kits, also offered individually for convenience
- Monarch Buffers and kits are designed with significantly less plastic than leading suppliers
- Désignation:Monarch® Buffer B3
- Taille:110 ml