Epic Basic Protection Coveralls
Latex free/low lint coverall for use in some critical/clean environments, and many industrial applications. Non-woven fabric is cloth-like and cool to wear. Features include a zipper front entry and elastic wrists.
- Cleanroom compatible design: Engineered for controlled environments to minimize particle shedding
- Basic protection level: Provides essential barrier against non-hazardous particulates and light liquid splashes
- Durable, low-linting fabric: Helps maintain cleanroom integrity and reduces contamination risk
- Full-body coverage: Includes long sleeves, full-length legs, and secure front closure
- Elastic cuffs: Ensures a snug fit and reduces particle escape
- Secure closure: Snap or zipper front for quick donning and doffing
- Available sizes: Multiple size options for proper fit across personnel
Full‑body, low‑lint protection for non‑critical cleanroom tasks.
The Tian's Epic Basic Protection Cleanroom Coverall delivers reliable particle barrier and comfort for routine operations in controlled environments. Made from durable, low‑linting fabric, it helps maintain cleanroom integrity while supporting operator mobility and all‑day wear. Featuring secure front closure, elasticized cuffs and ankles, and optional hood/boot integration (model‑dependent), this coverall is a cost‑effective choice for pharmaceutical, electronics, and medical device applications that require basic contamination control.
Garment is constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Benefits:
Contamination control: Maintains cleanliness standards in ISO class cleanrooms and controlled environments
Operator comfort: Lightweight, breathable material supports extended wear without discomfort
Cost efficiency: Ideal for non-critical processes, reducing unnecessary expense on higher-level garments
Compliance support: Meets cleanroom apparel requirements for GMP and ISO standards
Versatile use: Suitable for pharmaceutical, electronics, and medical device manufacturing
Ball burst: 82.58 N
Flammability: Class I
: Neatly folded and packed 25 pieces per case in a sealed inner poly-bag.
Latex free/low lint coverall for use in some critical/clean environments, and many industrial applications. Non-woven fabric is cloth-like and cool to wear. Features include a zipper front entry and elastic wrists.
- Cleanroom compatible design: Engineered for controlled environments to minimize particle shedding
- Basic protection level: Provides essential barrier against non-hazardous particulates and light liquid splashes
- Durable, low-linting fabric: Helps maintain cleanroom integrity and reduces contamination risk
- Full-body coverage: Includes long sleeves, full-length legs, and secure front closure
- Elastic cuffs: Ensures a snug fit and reduces particle escape
- Secure closure: Snap or zipper front for quick donning and doffing
- Available sizes: Multiple size options for proper fit across personnel
Full‑body, low‑lint protection for non‑critical cleanroom tasks.
The Tian's Epic Basic Protection Cleanroom Coverall delivers reliable particle barrier and comfort for routine operations in controlled environments. Made from durable, low‑linting fabric, it helps maintain cleanroom integrity while supporting operator mobility and all‑day wear. Featuring secure front closure, elasticized cuffs and ankles, and optional hood/boot integration (model‑dependent), this coverall is a cost‑effective choice for pharmaceutical, electronics, and medical device applications that require basic contamination control.
Garment is constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Benefits:
Contamination control: Maintains cleanliness standards in ISO class cleanrooms and controlled environments
Operator comfort: Lightweight, breathable material supports extended wear without discomfort
Cost efficiency: Ideal for non-critical processes, reducing unnecessary expense on higher-level garments
Compliance support: Meets cleanroom apparel requirements for GMP and ISO standards
Versatile use: Suitable for pharmaceutical, electronics, and medical device manufacturing
Ball burst: 82.58 N
Flammability: Class I
: Neatly folded and packed 25 pieces per case in a sealed inner poly-bag.
- Catalog No:
- 78150-484
- 78150-794
- 78150-472
- 78150-548
- 78150-778
- 78150-796
- 78150-664
- 78150-584
- 78150-786
- 78150-842
- 78150-790
- 78150-668
- 78150-496
- 78150-738
- 78150-504
- 78150-488
- 78150-848
- Fabric:
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Spunbound Polypropylene
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Level II
- Water resistance:
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- No Resistance
- Water vapor transmission rate:
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- 6856 g/m²/24 h
- Tensile strength:
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- 16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)
- Tearing strength:
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- 3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)
- Weight:
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
- 30 g/m²
Product Family Options
Coveralls w/elastic wrist and open ankle
- ColorTaille de confectionPackaging
- : 210881-2XLNew Product
Specifications
Cat. No.78150-484FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210881-3XLNew Product
Specifications
Cat. No.78150-794FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210881-LNew Product
Specifications
Cat. No.78150-472FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210881-MNew Product
Specifications
Cat. No.78150-548FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210881-SNew Product
Specifications
Cat. No.78150-778FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210881-XLNew Product
Specifications
Cat. No.78150-796FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product
Coveralls w/elastic wrist and ankle
- ColorTaille de confectionPackaging
- : 210882-2XLNew Product
Specifications
Cat. No.78150-664FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210882-3XLNew Product
Specifications
Cat. No.78150-584FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210882-4XLNew Product
Specifications
Cat. No.78150-786FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210882-LNew Product
Specifications
Cat. No.78150-842FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 210882-XLNew Product
Specifications
Cat. No.78150-790FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product
Coveralls w/attached hood and boot
- ColorTaille de confectionPackaging
- : 200881-2XLNew Product
Specifications
Cat. No.78150-668FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 200881-3XLNew Product
Specifications
Cat. No.78150-496FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 200881-4XLNew Product
Specifications
Cat. No.78150-738FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 200881-5XLNew Product
Specifications
Cat. No.78150-504FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 200881-LNew Product
Specifications
Cat. No.78150-488FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product - : 200881-XLNew Product
Specifications
Cat. No.78150-848FabricSpunbound PolypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroomParticle shedding (Helmke Drum)Level IIWater resistanceNo ResistanceWater vapor transmission rate6856 g/m²/24 hTensile strength16 lbs./inch (machine direction) 11 lbs./in (Cross Direction)Tearing strength3.5 lbs./inch (machine direction) 7.1 lbs./inch (cross direction)Weight30 g/m²New Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



