Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Conical Reaction Tubes

Conical Reaction Tubes

Conical Reaction Tubes
Supplier:  Greiner Bio-One
Certificates
Conical Reaction Tubes
Conical Reaction Tubes

2 Options Available Below

Product Details & Documents

Conical PP tube with blue screw cap, autoclavable.

  • High performance and durability - Conical 5 ml polypropylene tubes withstand –80 to +121 °C and up to 25000×g centrifugation

  • Secure and reliable - Blue screw caps ensure leak-proof sealing; Autoclavable for repeated use
  • Purity options - Available in sterile and non sterile versions to meet diverse lab requirements

Designed for precision and reliability, these conical polypropylene tubes deliver exceptional performance in demanding laboratory environments. Both versions feature a secure blue screw cap for leak-proof sealing. Resistant to extreme temperatures (–80 to +121 °C), they ensure durability and chemical stability. Ideal for sample preparation, storage, and transport in life science research, these tubes combine quality, versatility, and safety to meet the highest laboratory standards.

Documents

Product Family Options

Product Information

  • Désignation
    Capacité
    Matière
    Color
    Details
    Cap Color
    Cap Type
    Packaging
    Sterility
    Availability
    Price
  • Conical reaction tubes
    5 ml
    Polypropylene
    Clear
    DNase/RNase/DNA/pyrogen-free
    Blue
    Screw cap
    100 tubes per bag
    Non sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $238.26
    $238.26
    New Customer Promotional Price
    Case of 500
       $238.26

    Supplier Number: 725201
  • Conical reaction tubes
    5 ml
    Polypropylene
    Clear
    DNase/RNase/DNA/pyrogen-free
    Blue
    Screw cap
    50 tubes per rack
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $398.33
    $398.33
    New Customer Promotional Price
    Case of 500
       $398.33

    Supplier Number: 725261
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.