Transferpette® pro, Ejector (bottom part)
New Product
Supplier: Brandtech
Jump to:
À propos de cet article
Replacement lower ejector.
Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
9 Options Available Below
Product Details & Documents
Replacement lower ejector.
Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
Product Family Options
Informations sur le produit
- VolumeAvailabilityPrice
- Supplier Number: 704712Spécifications:New ProductNew Product
- Supplier Number: 704705Spécifications:New ProductNew Product
- Supplier Number: 704706Spécifications:New ProductNew Product
- Supplier Number: 704707Spécifications:New ProductNew Product
- Supplier Number: 704710Spécifications:New ProductNew Product
- Supplier Number: 704711Spécifications:New ProductNew Product
- Supplier Number: 704713Spécifications:New ProductNew Product
- Supplier Number: 704714Spécifications:New ProductNew Product
- Supplier Number: 704715Spécifications:New ProductNew Product