Transferpette® pro, Ejector Set (upper and lower)
New Product
Supplier: Brandtech
Jump to:
À propos de cet article
Replacement ejector set.
Ejector set (upper and lower part) for use with Transferpette® pro single channel pipettors, 2.5 to 10ml
Designed for a perfect fit, simple installation, and long-lasting reliability.
3 Options Available Below
Product Details & Documents
Replacement ejector set.
Ejector set (upper and lower part) for use with Transferpette® pro single channel pipettors, 2.5 to 10ml
Designed for a perfect fit, simple installation, and long-lasting reliability.
Product Family Options
Informations sur le produit
- VolumeAvailabilityPrice
- Supplier Number: 704757Spécifications:New ProductNew Product
- Supplier Number: 704755Spécifications:New ProductNew Product
- Supplier Number: 704756Spécifications:New ProductNew Product