Adhesive Foil Seal
Adhesive Foil Seal
Adhesive foil seal, Non sterile
Catalog # 76808-526
Supplier #:  899405-1
Adhesive Foil Seal
Catalog # 76808-526
Adhesive Foil Seal
Adhesive Foil Seal
Adhesive foil seal, Non sterile
Price
$152.00
Log in for availability
Case of 100
Minimum order quantity is 1
Product Details & Documents
Thomson Instrument Company adhesive foil seals are for short and long-term storage in well plates.
  • For use with all plates
Thomson well plates are offered in 24- and 96-well configurations with a V, U or pyramid shape bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Many plates are individually wrapped, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Specifications
  • Sterility:
    Non sterile
  • Désignation:
    Adhesive foil seal