Filter Plates
Filter Plates
Certificates
Filter Plates
7 Options Available Below
Product Details & Documents
Thomson filter plates (24- and 96-well) enable fast, parallel plate-to-plate processing for biological and analytical workflows. For clarification, the Universal Rapid Clear® 24-well depth-filtration plate is automation-compatible and clarifies cell-culture supernatants or lysates in ~5 minutes by centrifuge. The graded, multi-layer stack removes whole cells and debris, including lysis byproducts, while minimizing interaction with target proteins. Pair with Thomson collection plates and seals for clean transfers and high-throughput screening.
  • Universal Rapid Clear® depth filtration; multi-layer, graded matrix captures progressively smaller cells, debris, and aggregates for reliable clarification by swing-bucket centrifuge
  • Filtration membranes in PTFE/PVDF options support final filtration and solvent-compatible analytical steps
  • Purification option: 25 µm polypropylene for resin/adsorption workflows
  • Square well/long drip well shapes
Stack a Thomson filter plate over a collection plate to process 24- or 96-well sample sets under consistent conditions. The lineup spans clarification (Universal Rapid Clear®), filtration (PTFE/PVDF), and purification (25 µm PP), matched to centrifuge, vacuum, or positive-pressure processing. This integrated setup streamlines workflows, reduces manual steps, and improves reproducibility for high-throughput screening.
Product Family Options
Informations sur le produit
  • Désignation
    Nb de puits
    Profil
    Jupe
    Well Volume
    Availability
    Price
  • Rapid Clear® 0.2 µm filter plate, SAL 10⁻⁶ (Compatible with collection plate 931565-G-1X)
    24
    Standard
    Fully skirted
    9 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $2,470.00
    $2,470.00
    Case of 20
       $2,470.00
    Supplier Number: 921546
  • Filter plate, 25 µm polypropylene, non sterile (Compatible with collection plate 931130)
    96
    Standard
    Fully skirted
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $1,660.00
    $1,660.00
    Case of 25
       $1,660.00
    Supplier Number: 931919
  • Filter plate, 25 µm polypropylene, non sterile (Compatible with collection plate 931565-G-1X)
    24
    Standard
    Fully skirted
    10.8 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $1,660.00
    $1,660.00
    Case of 20
       $1,660.00
    Supplier Number: 921550
  • Filter plate, 0.2 µm PTFE, non sterile (Compatible with collection plate 931130)
    96
    Standard
    Fully skirted
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $1,320.00
    $1,320.00
    Case of 20
       $1,320.00
    Supplier Number: 921730
  • Filter plate, 0.2 µm PVDF, non sterile (Compatible with collection plate 931130)
    96
    Standard
    Fully skirted
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $1,320.00
    $1,320.00
    Case of 20
       $1,320.00
    Supplier Number: 921731
  • Filter plate, 0.45 µm PTFE, non sterile (Compatible with collection plate 931130)
    96
    Standard
    Fully skirted
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $1,320.00
    $1,320.00
    Case of 20
       $1,320.00
    Supplier Number: 921740
  • Rapid Clear® 0.2 µm filter plate, SAL 10⁻⁶ (Compatible with collection plate 931130)
    96
    Standard
    Fully skirted
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $4,220.00
    $4,220.00
    Case of 20
       $4,220.00
    Supplier Number: 921746
Accessories
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Pierceable foil heat seal
    Pierceable foil heat seal
    76808-538
    Non sterile
    Log in pour la disponibilité
    Case of 100 - $225.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum manifold
    Vacuum manifold
    76808-542
    Log in pour la disponibilité
    Each - $4,760.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Well plate lid
    Well plate lid
    76808-458
    Non sterile
    Log in pour la disponibilité
    Case of 100 - $343.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Airporous plate seal
    Airporous plate seal
    76808-536
    Sterile
    Log in pour la disponibilité
    Case of 100 - $230.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • 96-well capmat, round well
    96-well capmat, round well
    76808-356
    Non sterile
    Log in pour la disponibilité
    Case of 50 - $609.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • 96-well capmat, square well
    96-well capmat, square well
    76808-358
    Non sterile
    Log in pour la disponibilité
    Case of 100 - $547.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Adhesive foil seal
    Adhesive foil seal
    76808-526
    Non sterile
    Log in pour la disponibilité
    Case of 100 - $152.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Foil heat seal for long-term storage
    Foil heat seal for long-term storage
    76808-540
    Non sterile
    Log in pour la disponibilité
    Case of 100 - $225.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® integrated lid for 6-well plates
    Optimum Growth® integrated lid for 6-well plates
    77971-810
    Sterile
    Log in pour la disponibilité
    Case of 20 - $605.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Well plate lid
    Well plate lid
    76808-456
    Sterile
    Log in pour la disponibilité
    Case of 100 - $464.00