Optifit Pipette Tips
New Product
 :   Sartorius
Optifit Pipette Tips
 :  Sartorius
New Product
Optifit Pipette Tips
Optifit Pipette Tips
The non-filtered Optifit tips are designed to seamlessly fit together with Sartorius pipettes. For applications involving larger volume transfers, the 10 ml Optifit tips are specifically designed to deliver precise and accurate performance.
  • Informative rack labels indicating tip volume, product number, lot number, expiration date, and purity level for improved tip identification and traceability
The Optifit tips are intended, designed and manufactured to be used in liquid dispensing in a variety of applications and to be used in combination with Sartorius pipettes.
Product Family Options
  • Volume
    Longueur
    Tip Color
    Color Code
    For Use With
    Packed
  • 10 ml
    164 mm
    Transparent
    Red
    Sartorius pipettes
    Pack with 4 boxes, each box 35 tips
    Pack of 140
    Pack of 140 - $192.00
    $192.00
    Pack of 140
       $192.00
     : LH-799000
    New Product
    New Product
  • 10 ml
    164 mm
    Transparent
    Red
    Sartorius pipettes
    1 bag
    Pack of 210
    Pack of 210 - $76.70
    $76.70
    Pack of 210
       $76.70
     : LH-799004