Ultem® Socket Cap
Ultem® Socket cap, PEI
78474-433
New Product
: SAVILLEX
Supplier #: 730-5132
Socket-type cap is molded of Ultem® for extra strength. Slips over PFA closure for additional holding in pressure applications.
Service temperature to 135 °C (275 °F)
Allows greater torque to be applied to square body vials for sealing
- Désignation:Ultem® Socket cap
- Matière:PEI