Instant Explosive Detection Wipes
About this item
Trace Eye-D instant explosives detection wipes provide rapid, field-ready identification of trace explosive residues, including TNT, PETN, RDX, TATP, ammonium nitrate, chlorates, and perchlorates. They require no specialized equipment and use simple color change chemistry for safe on-scene screening.
- Rapid detection results of trace explosive residues
- Field-ready packaging for on-scene operations
- Trusted globally by law enforcement, corrections, and security teams
- Simple color change results; no instruments, mixing, or glass ampoules
- Works on solids, liquids, packaging, and surfaces
- Minimizes exposure with safe stand-off sampling
- Compact, lightweight, and easy to carry in any kit
- Durable 6×3" polyester towelette allows refolding for testing several areas with one wipe
- Negative result can be disposed of in common waste stream
- Made in the USA
Trace Eye-D instant explosives detection wipes are simple to use, and require no specialized equipment. They employ color change chemistry for fast on-scene screening; just open, wipe, and look. Each wipe features a large 6×3" 100% polyester towelette, refoldable to test multiple surfaces, and is housed in a durable 4×3" multi-layer pouch (foil, LDPE, polyester, EAA) weighing 2.54 g total. Trace Eye-D wipes are pretreated, and must be used within 30 minutes after opening. They should be used within one year of purchase for optimal results. Trace Eye-D wipes are unaffected by heat, cold, or altitude and are lightweight and portable, making them ideal for law enforcement, military, aviation security, corrections, K-9, and bomb squad personnel.
Chemical reaction attributes: Aliphatic explosives detection wipes utilize Griess. Chlorate-based explosives detection wipes utilize REDOX. Aromatic explosives detection wipes utilize Meisenheimer Complex Reaction/Formation. Peroxide-based explosives detection wipes utilize REDOX. Perchlorate-based Oxidizer detection wipes utilize Precipitation Reaction.
Certifications: Berry compliant.
Packaging: Resealable bag contains 10 individually wrapped wipes; bulk options are loose indivudually wrapped wipes (no outer resealable packaging).
12 Options Available Below
Product Details & Documents
Trace Eye-D instant explosives detection wipes provide rapid, field-ready identification of trace explosive residues, including TNT, PETN, RDX, TATP, ammonium nitrate, chlorates, and perchlorates. They require no specialized equipment and use simple color change chemistry for safe on-scene screening.
- Rapid detection results of trace explosive residues
- Field-ready packaging for on-scene operations
- Trusted globally by law enforcement, corrections, and security teams
- Simple color change results; no instruments, mixing, or glass ampoules
- Works on solids, liquids, packaging, and surfaces
- Minimizes exposure with safe stand-off sampling
- Compact, lightweight, and easy to carry in any kit
- Durable 6×3" polyester towelette allows refolding for testing several areas with one wipe
- Negative result can be disposed of in common waste stream
- Made in the USA
Trace Eye-D instant explosives detection wipes are simple to use, and require no specialized equipment. They employ color change chemistry for fast on-scene screening; just open, wipe, and look. Each wipe features a large 6×3" 100% polyester towelette, refoldable to test multiple surfaces, and is housed in a durable 4×3" multi-layer pouch (foil, LDPE, polyester, EAA) weighing 2.54 g total. Trace Eye-D wipes are pretreated, and must be used within 30 minutes after opening. They should be used within one year of purchase for optimal results. Trace Eye-D wipes are unaffected by heat, cold, or altitude and are lightweight and portable, making them ideal for law enforcement, military, aviation security, corrections, K-9, and bomb squad personnel.
Chemical reaction attributes: Aliphatic explosives detection wipes utilize Griess. Chlorate-based explosives detection wipes utilize REDOX. Aromatic explosives detection wipes utilize Meisenheimer Complex Reaction/Formation. Peroxide-based explosives detection wipes utilize REDOX. Perchlorate-based Oxidizer detection wipes utilize Precipitation Reaction.
Certifications: Berry compliant.
Packaging: Resealable bag contains 10 individually wrapped wipes; bulk options are loose indivudually wrapped wipes (no outer resealable packaging).
Product Family Options
Aliphatic Explosives Detection
Product Information
- DésignationAvailabilityPrice
- Target substances: Penaerythitol Tetranitrate (PETN), Erythritol Tetranitrate (ETN), Cyclotrimethylenetrinitramine (RDX), Cyclotretramethylene Tetranitramine (HMX), Composition-4 (C-4), Nitrocellulose (NC, Guncotton), Urea Nitrate (UN), ANFO (AN/FO), Ammonium Nitrate (AN)Log in for availabilityPack of 50$414.73Supplier Number: 948216Specifications:New Product
More
Specifications
Cat. No.78568-288New Product - Target substances: Penaerythitol Tetranitrate (PETN), Erythritol Tetranitrate (ETN), Cyclotrimethylenetrinitramine (RDX), Cyclotretramethylene Tetranitramine (HMX), Composition-4 (C-4), Nitrocellulose (NC, Guncotton), Urea Nitrate (UN), ANFO (AN/FO), Ammonium Nitrate (AN)Log in for availabilityPack of 100$805.78Supplier Number: 948223Specifications:New Product
More
Specifications
Cat. No.78568-280New Product - Target substances: Penaerythitol Tetranitrate (PETN), Erythritol Tetranitrate (ETN), Cyclotrimethylenetrinitramine (RDX), Cyclotretramethylene Tetranitramine (HMX), Composition-4 (C-4), Nitrocellulose (NC, Guncotton), Urea Nitrate (UN), ANFO (AN/FO), Ammonium Nitrate (AN)Log in for availabilityPack of 10$86.88Supplier Number: 948070Specifications:New Product
More
Specifications
Cat. No.78568-272New Product
Chlorate-based Explosives Detection
Product Information
- DésignationAvailabilityPrice
- Supplier Number: 948230Specifications:New Product
More
Specifications
Cat. No.78568-266New Product - Supplier Number: 948247Specifications:New Product
More
Specifications
Cat. No.78568-292New Product - Supplier Number: 948056Specifications:New Product
More
Specifications
Cat. No.78568-294New Product
Aromatic Explosives Detection
Product Information
- DésignationAvailabilityPrice
- Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)Log in for availabilityPack of 10$86.88Supplier Number: 948049Specifications:New Product
More
Specifications
Cat. No.78568-274New Product - Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)Log in for availabilityPack of 50$414.73Supplier Number: 948254Specifications:New Product
More
Specifications
Cat. No.78568-310New Product - Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)Log in for availabilityPack of 100$805.78Supplier Number: 948261Specifications:New Product
More
Specifications
Cat. No.78568-268New Product
Perchlorate-based Oxidizer Detection
Product Information
- DésignationAvailabilityPrice
- Target substances: Ammonium Perchlorate, Potassium PerchlorateLog in for availabilityPack of 10$86.88Supplier Number: 948063Specifications:New Product
More
Specifications
Cat. No.78568-270New Product - Target substances: Ammonium Perchlorate, Potassium PerchlorateLog in for availabilityPack of 50$414.73Supplier Number: 948292Specifications:New Product
More
Specifications
Cat. No.78568-276New Product - Target substances: Ammonium Perchlorate, Potassium PerchlorateLog in for availabilityPack of 100$805.78Supplier Number: 948308Specifications:New Product
More
Specifications
Cat. No.78568-300New Product