Instant Explosive Detection Wipes
Instant Explosive Detection Wipes
New Product
Supplier:   TRACE EYE D LLC
Instant Explosive Detection Wipes
Supplier:  TRACE EYE D LLC
New Product
Certificates
Instant Explosive Detection Wipes
Instant Explosive Detection Wipes
12 Options Available Below
Product Details & Documents
Trace Eye-D instant explosives detection wipes provide rapid, field-ready identification of trace explosive residues, including TNT, PETN, RDX, TATP, ammonium nitrate, chlorates, and perchlorates. They require no specialized equipment and use simple color change chemistry for safe on-scene screening.
  • Rapid detection results of trace explosive residues
  • Field-ready packaging for on-scene operations
  • Trusted globally by law enforcement, corrections, and security teams
  • Simple color change results; no instruments, mixing, or glass ampoules
  • Works on solids, liquids, packaging, and surfaces
  • Minimizes exposure with safe stand-off sampling
  • Compact, lightweight, and easy to carry in any kit
  • Durable 6×3" polyester towelette allows refolding for testing several areas with one wipe
  • Negative result can be disposed of in common waste stream
  • Made in the USA
Trace Eye-D instant explosives detection wipes are simple to use, and require no specialized equipment. They employ color change chemistry for fast on-scene screening; just open, wipe, and look. Each wipe features a large 6×3" 100% polyester towelette, refoldable to test multiple surfaces, and is housed in a durable 4×3" multi-layer pouch (foil, LDPE, polyester, EAA) weighing 2.54 g total. Trace Eye-D wipes are pretreated, and must be used within 30 minutes after opening. They should be used within one year of purchase for optimal results. Trace Eye-D wipes are unaffected by heat, cold, or altitude and are lightweight and portable, making them ideal for law enforcement, military, aviation security, corrections, K-9, and bomb squad personnel.
Chemical reaction attributes: Aliphatic explosives detection wipes utilize Griess. Chlorate-based explosives detection wipes utilize REDOX. Aromatic explosives detection wipes utilize Meisenheimer Complex Reaction/Formation. Peroxide-based explosives detection wipes utilize REDOX. Perchlorate-based Oxidizer detection wipes utilize Precipitation Reaction.
Certifications: Berry compliant.
Packaging: Resealable bag contains 10 individually wrapped wipes; bulk options are loose indivudually wrapped wipes (no outer resealable packaging).
Product Family Options
Aliphatic Explosives Detection
Product Information
  • Désignation
    Availability
    Price
  • Target substances: Penaerythitol Tetranitrate (PETN), Erythritol Tetranitrate (ETN), Cyclotrimethylenetrinitramine (RDX), Cyclotretramethylene Tetranitramine (HMX), Composition-4 (C-4), Nitrocellulose (NC, Guncotton), Urea Nitrate (UN), ANFO (AN/FO), Ammonium Nitrate (AN)
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $414.73
    $414.73
    Pack of 50
       $414.73
    Minimum order quantity is 1
    Supplier Number: 948216
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-288
    New Product
  • Target substances: Penaerythitol Tetranitrate (PETN), Erythritol Tetranitrate (ETN), Cyclotrimethylenetrinitramine (RDX), Cyclotretramethylene Tetranitramine (HMX), Composition-4 (C-4), Nitrocellulose (NC, Guncotton), Urea Nitrate (UN), ANFO (AN/FO), Ammonium Nitrate (AN)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $805.78
    $805.78
    Pack of 100
       $805.78
    Minimum order quantity is 1
    Supplier Number: 948223
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-280
    New Product
  • Target substances: Penaerythitol Tetranitrate (PETN), Erythritol Tetranitrate (ETN), Cyclotrimethylenetrinitramine (RDX), Cyclotretramethylene Tetranitramine (HMX), Composition-4 (C-4), Nitrocellulose (NC, Guncotton), Urea Nitrate (UN), ANFO (AN/FO), Ammonium Nitrate (AN)
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $86.88
    $86.88
    Pack of 10
       $86.88
    Minimum order quantity is 1
    Supplier Number: 948070
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-272
    New Product
Chlorate-based Explosives Detection
Product Information
  • Désignation
    Availability
    Price
  • Target substances: Ammonium Chlorate, Potassium Chlorate
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $414.73
    $414.73
    Pack of 50
       $414.73
    Minimum order quantity is 1
    Supplier Number: 948230
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-266
    New Product
  • Target substances: Ammonium Chlorate, Potassium Chlorate
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $805.78
    $805.78
    Pack of 100
       $805.78
    Minimum order quantity is 1
    Supplier Number: 948247
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-292
    New Product
  • Target substances: Ammonium Chlorate, Potassium Chlorate
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $86.88
    $86.88
    Pack of 10
       $86.88
    Minimum order quantity is 1
    Supplier Number: 948056
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-294
    New Product
Aromatic Explosives Detection
Product Information
  • Désignation
    Availability
    Price
  • Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $86.88
    $86.88
    Pack of 10
       $86.88
    Minimum order quantity is 1
    Supplier Number: 948049
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-274
    New Product
  • Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $414.73
    $414.73
    Pack of 50
       $414.73
    Minimum order quantity is 1
    Supplier Number: 948254
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-310
    New Product
  • Target substances: 2,4-Dinitrotoluene (DNT), 2,6-Dinitrotoluene (DNT), Trinitrotoluene (TNT)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $805.78
    $805.78
    Pack of 100
       $805.78
    Minimum order quantity is 1
    Supplier Number: 948261
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-268
    New Product
Perchlorate-based Oxidizer Detection
Product Information
  • Désignation
    Availability
    Price
  • Target substances: Ammonium Perchlorate, Potassium Perchlorate
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $86.88
    $86.88
    Pack of 10
       $86.88
    Minimum order quantity is 1
    Supplier Number: 948063
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-270
    New Product
  • Target substances: Ammonium Perchlorate, Potassium Perchlorate
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $414.73
    $414.73
    Pack of 50
       $414.73
    Minimum order quantity is 1
    Supplier Number: 948292
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-276
    New Product
  • Target substances: Ammonium Perchlorate, Potassium Perchlorate
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $805.78
    $805.78
    Pack of 100
       $805.78
    Minimum order quantity is 1
    Supplier Number: 948308
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78568-300
    New Product